Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 180984511-180984784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC19
Entrez Gene ID
131118
Gene Name
DnaJ heat shock protein family (Hsp40) member C19
Gene Aliases
PAM18, TIM14, TIMM14
UniGene
Hs.230601
Gene Chromosome Location
Chr.3: 180983709-180989774 on Build GRCh38
Transcripts
NM_001190233.1 NM_145261.3 NR_033721.1 NR_033722.1 NR_033723.1 NR_046073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1421144 c.348delA p.K116fs? Chr.3: 180984643-180984643 on Build GRCh38
3408457 c.316G>A p.A106T Chr.3: 180984675-180984675 on Build GRCh38
446047 c.325T>C p.L109L Chr.3: 180984666-180984666 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753816728 Chr.3: 180984561-180984561 on Build GRCh38
rs763986833 Chr.3: 180984657-180984659 on Build GRCh38
rs764876279 Chr.3: 180984592-180984592 on Build GRCh38
rs771066032 Chr.3: 180984544-180984544 on Build GRCh38
rs775210714 Chr.3: 180984597-180984600 on Build GRCh38
rs754864030 Chr.3: 180984721-180984721 on Build GRCh38
rs1043123757 Chr.3: 180984625-180984625 on Build GRCh38
rs771098271 Chr.3: 180984643-180984643 on Build GRCh38
rs370042988 Chr.3: 180984660-180984660 on Build GRCh38
rs771901822 Chr.3: 180984580-180984580 on Build GRCh38
rs151056679 Chr.3: 180984710-180984710 on Build GRCh38
rs766348961 Chr.3: 180984658-180984658 on Build GRCh38
rs761771242 Chr.3: 180984586-180984586 on Build GRCh38
rs968957502 Chr.3: 180984659-180984659 on Build GRCh38
rs535208857 Chr.3: 180984564-180984564 on Build GRCh38
rs764813798 Chr.3: 180984724-180984725 on Build GRCh38
rs150673364 Chr.3: 180984734-180984734 on Build GRCh38
rs763026371 Chr.3: 180984536-180984536 on Build GRCh38
rs761342359 Chr.3: 180984695-180984696 on Build GRCh38
rs587777224 Chr.3: 180984691-180984691 on Build GRCh38
rs373886730 Chr.3: 180984691-180984691 on Build GRCh38
rs547217176 Chr.3: 180984601-180984601 on Build GRCh38
rs3026210 Chr.3: 180984725-180984725 on Build GRCh38
rs17850540 Chr.3: 180984706-180984706 on Build GRCh38
rs781713901 Chr.3: 180984617-180984617 on Build GRCh38
rs774534561 Chr.3: 180984549-180984549 on Build GRCh38
rs759658546 Chr.3: 180984555-180984555 on Build GRCh38
rs372375155 Chr.3: 180984612-180984612 on Build GRCh38
rs766954639 Chr.3: 180984715-180984715 on Build GRCh38
rs750082106 Chr.3: 180984733-180984734 on Build GRCh38
rs141982712 Chr.3: 180984722-180984722 on Build GRCh38
rs200732155 Chr.3: 180984665-180984665 on Build GRCh38
rs944295139 Chr.3: 180984596-180984596 on Build GRCh38
rs536157013 Chr.3: 180984613-180984613 on Build GRCh38
rs1023070337 Chr.3: 180984628-180984628 on Build GRCh38
rs776716974 Chr.3: 180984661-180984661 on Build GRCh38
rs748585514 Chr.3: 180984621-180984621 on Build GRCh38
rs753224190 Chr.3: 180984716-180984716 on Build GRCh38
rs749557429 Chr.3: 180984638-180984638 on Build GRCh38
rs760648011 Chr.3: 180984603-180984606 on Build GRCh38
rs757628797 Chr.3: 180984755-180984755 on Build GRCh38
rs982925602 Chr.3: 180984579-180984579 on Build GRCh38
rs993200454 Chr.3: 180984595-180984595 on Build GRCh38
rs202185352 Chr.3: 180984603-180984603 on Build GRCh38
rs766074857 Chr.3: 180984705-180984705 on Build GRCh38
rs1037501435 Chr.3: 180984542-180984542 on Build GRCh38
rs764176644 Chr.3: 180984557-180984557 on Build GRCh38
rs762783413 Chr.3: 180984696-180984696 on Build GRCh38
rs756512724 Chr.3: 180984627-180984627 on Build GRCh38
rs777951711 Chr.3: 180984637-180984637 on Build GRCh38
rs201233020 Chr.3: 180984680-180984680 on Build GRCh38

View Full Product Details