Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70450112-70450385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG3
Entrez Gene ID
1741
Gene Name
discs large MAGUK scaffold protein 3
Gene Aliases
MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR
UniGene
Hs.721586
Gene Chromosome Location
Chr.X: 70444850-70505490 on Build GRCh38
Transcripts
NM_021120.3 XM_006724625.2 XM_006724626.2 XM_011530883.1

SNP Detail

SNP ID Chromosome Location
rs144839135 Chr.X: 70450253-70450253 on Build GRCh38
rs769596271 Chr.X: 70450314-70450314 on Build GRCh38
rs747133636 Chr.X: 70450346-70450346 on Build GRCh38
rs778537059 Chr.X: 70450170-70450170 on Build GRCh38
rs141106519 Chr.X: 70450251-70450251 on Build GRCh38
rs199927598 Chr.X: 70450276-70450276 on Build GRCh38
rs138759292 Chr.X: 70450296-70450296 on Build GRCh38
rs747584382 Chr.X: 70450195-70450195 on Build GRCh38
rs756603402 Chr.X: 70450163-70450163 on Build GRCh38
rs770784364 Chr.X: 70450279-70450279 on Build GRCh38
rs746816239 Chr.X: 70450258-70450258 on Build GRCh38
rs757563882 Chr.X: 70450141-70450141 on Build GRCh38
rs750859809 Chr.X: 70450162-70450162 on Build GRCh38
rs762898798 Chr.X: 70450320-70450320 on Build GRCh38
rs753531909 Chr.X: 70450317-70450317 on Build GRCh38
rs751981534 Chr.X: 70450139-70450139 on Build GRCh38
rs373430026 Chr.X: 70450145-70450145 on Build GRCh38
rs757999224 Chr.X: 70450351-70450351 on Build GRCh38
rs776562185 Chr.X: 70450281-70450281 on Build GRCh38
rs4844120 Chr.X: 70450331-70450331 on Build GRCh38

View Full Product Details