Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 80205514-80205787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIO2
Entrez Gene ID
1734
Gene Name
deiodinase, iodothyronine, type II
Gene Aliases
5DII, D2, DIOII, SelY, TXDI2
UniGene
Hs.202354
Gene Chromosome Location
Chr.14: 80197525-80231054 on Build GRCh38
Transcripts
NM_000793.5 NM_001007023.4 NM_001242502.2 NM_001242503.2 NM_001324462.1 NM_013989.4

SNP Detail

SNP ID Chromosome Location
rs772484692 Chr.14: 80205645-80205645 on Build GRCh38
rs745385180 Chr.14: 80205625-80205626 on Build GRCh38
rs767611570 Chr.14: 80205714-80205717 on Build GRCh38
rs766645776 Chr.14: 80205657-80205657 on Build GRCh38
rs1023059362 Chr.14: 80205604-80205604 on Build GRCh38
rs748654165 Chr.14: 80205632-80205632 on Build GRCh38
rs748959744 Chr.14: 80205624-80205624 on Build GRCh38
rs565586477 Chr.14: 80205744-80205744 on Build GRCh38
rs749866953 Chr.14: 80205734-80205734 on Build GRCh38
rs776993197 Chr.14: 80205664-80205664 on Build GRCh38
rs758926851 Chr.14: 80205634-80205635 on Build GRCh38
rs750718171 Chr.14: 80205688-80205688 on Build GRCh38
rs765536454 Chr.14: 80205680-80205680 on Build GRCh38
rs758722588 Chr.14: 80205737-80205737 on Build GRCh38
rs941684857 Chr.14: 80205541-80205541 on Build GRCh38
rs769049241 Chr.14: 80205630-80205630 on Build GRCh38
rs77075531 Chr.14: 80205601-80205601 on Build GRCh38
rs745403963 Chr.14: 80205587-80205587 on Build GRCh38
rs907471294 Chr.14: 80205676-80205676 on Build GRCh38
rs757180288 Chr.14: 80205700-80205700 on Build GRCh38
rs748944597 Chr.14: 80205760-80205760 on Build GRCh38
rs773379888 Chr.14: 80205651-80205651 on Build GRCh38
rs780650637 Chr.14: 80205583-80205583 on Build GRCh38
rs368731809 Chr.14: 80205726-80205726 on Build GRCh38
rs1028408073 Chr.14: 80205595-80205595 on Build GRCh38
rs35983132 Chr.14: 80205562-80205563 on Build GRCh38
rs769378214 Chr.14: 80205599-80205599 on Build GRCh38
rs780740869 Chr.14: 80205685-80205686 on Build GRCh38
rs757419780 Chr.14: 80205570-80205570 on Build GRCh38
rs760853066 Chr.14: 80205654-80205654 on Build GRCh38
rs548845517 Chr.14: 80205668-80205668 on Build GRCh38
rs990540128 Chr.14: 80205703-80205703 on Build GRCh38
rs562369066 Chr.14: 80205627-80205627 on Build GRCh38

View Full Product Details