Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 148614189-148614414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf33
Entrez Gene ID
202865
Gene Name
chromosome 7 open reading frame 33
Gene Aliases
-
UniGene
Hs.352212
Gene Chromosome Location
Chr.7: 148590565-148615860 on Build GRCh38
Transcripts
NM_145304.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
243404 c.445A>T p.S149C Chr.7: 148614282-148614282 on Build GRCh38
6176710 c.443C>G p.T148R Chr.7: 148614280-148614280 on Build GRCh38
3635727 c.357C>T p.G119G Chr.7: 148614194-148614194 on Build GRCh38
4495245 c.449C>T p.S150L Chr.7: 148614286-148614286 on Build GRCh38
5948165 c.459+1G>A p.? Chr.7: 148614297-148614297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780114609 Chr.7: 148614292-148614292 on Build GRCh38
rs765616367 Chr.7: 148614377-148614377 on Build GRCh38
rs758306046 Chr.7: 148614260-148614260 on Build GRCh38
rs776187715 Chr.7: 148614317-148614317 on Build GRCh38
rs201586129 Chr.7: 148614216-148614216 on Build GRCh38
rs752532113 Chr.7: 148614258-148614258 on Build GRCh38
rs368502957 Chr.7: 148614320-148614320 on Build GRCh38
rs749428289 Chr.7: 148614295-148614295 on Build GRCh38
rs372717391 Chr.7: 148614336-148614336 on Build GRCh38
rs746914805 Chr.7: 148614281-148614281 on Build GRCh38
rs773484418 Chr.7: 148614306-148614306 on Build GRCh38
rs113864617 Chr.7: 148614304-148614304 on Build GRCh38
rs200645489 Chr.7: 148614323-148614323 on Build GRCh38
rs772381621 Chr.7: 148614305-148614305 on Build GRCh38
rs764056902 Chr.7: 148614249-148614250 on Build GRCh38
rs577343864 Chr.7: 148614313-148614313 on Build GRCh38
rs909640056 Chr.7: 148614280-148614280 on Build GRCh38
rs754614935 Chr.7: 148614221-148614221 on Build GRCh38
rs534493236 Chr.7: 148614220-148614220 on Build GRCh38
rs768863009 Chr.7: 148614296-148614296 on Build GRCh38
rs761181588 Chr.7: 148614309-148614309 on Build GRCh38
rs188854544 Chr.7: 148614367-148614367 on Build GRCh38
rs952897681 Chr.7: 148614365-148614365 on Build GRCh38
rs141295114 Chr.7: 148614297-148614297 on Build GRCh38
rs145413433 Chr.7: 148614283-148614283 on Build GRCh38
rs778762571 Chr.7: 148614240-148614240 on Build GRCh38
rs865881632 Chr.7: 148614248-148614249 on Build GRCh38
rs199640449 Chr.7: 148614215-148614215 on Build GRCh38
rs1051976161 Chr.7: 148614311-148614311 on Build GRCh38
rs112219553 Chr.7: 148614274-148614274 on Build GRCh38

View Full Product Details