Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2360707-2360938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3B
Entrez Gene ID
8662
Gene Name
eukaryotic translation initiation factor 3 subunit B
Gene Aliases
EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, PRT1
UniGene
Hs.371001
Gene Chromosome Location
Chr.7: 2354540-2380745 on Build GRCh38
Transcripts
NM_001037283.1 NM_003751.3 XM_011515599.1 XM_011515600.1 XM_011515601.1 XM_017012752.1 XM_017012753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1698323 c.526C>T p.R176W Chr.7: 2360736-2360736 on Build GRCh38
136588 c.535G>A p.E179K Chr.7: 2360745-2360745 on Build GRCh38
7153850 c.643G>C p.G215R Chr.7: 2360853-2360853 on Build GRCh38
3778337 c.596G>A p.R199Q Chr.7: 2360806-2360806 on Build GRCh38
6652800 c.646A>G p.K216E Chr.7: 2360856-2360856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147152815 Chr.7: 2360792-2360792 on Build GRCh38
rs775088394 Chr.7: 2360880-2360880 on Build GRCh38
rs201331895 Chr.7: 2360737-2360737 on Build GRCh38
rs993755036 Chr.7: 2360741-2360741 on Build GRCh38
rs375454344 Chr.7: 2360912-2360912 on Build GRCh38
rs374070548 Chr.7: 2360765-2360765 on Build GRCh38
rs764593785 Chr.7: 2360908-2360908 on Build GRCh38
rs769911546 Chr.7: 2360758-2360758 on Build GRCh38
rs764191350 Chr.7: 2360797-2360797 on Build GRCh38
rs922864560 Chr.7: 2360886-2360886 on Build GRCh38
rs750629541 Chr.7: 2360789-2360789 on Build GRCh38
rs201155310 Chr.7: 2360766-2360766 on Build GRCh38
rs770621899 Chr.7: 2360828-2360828 on Build GRCh38
rs563978553 Chr.7: 2360820-2360820 on Build GRCh38
rs372472481 Chr.7: 2360780-2360780 on Build GRCh38
rs762297094 Chr.7: 2360913-2360913 on Build GRCh38
rs773012336 Chr.7: 2360744-2360744 on Build GRCh38
rs377134404 Chr.7: 2360783-2360783 on Build GRCh38
rs748107158 Chr.7: 2360799-2360799 on Build GRCh38
rs769457046 Chr.7: 2360865-2360865 on Build GRCh38
rs58174334 Chr.7: 2360759-2360759 on Build GRCh38
rs143426217 Chr.7: 2360782-2360782 on Build GRCh38
rs745482471 Chr.7: 2360862-2360862 on Build GRCh38
rs780069446 Chr.7: 2360795-2360795 on Build GRCh38
rs746890634 Chr.7: 2360802-2360802 on Build GRCh38
rs776398952 Chr.7: 2360858-2360858 on Build GRCh38
rs748047621 Chr.7: 2360736-2360736 on Build GRCh38
rs746875139 Chr.7: 2360779-2360779 on Build GRCh38
rs771989737 Chr.7: 2360800-2360800 on Build GRCh38
rs757423281 Chr.7: 2360786-2360786 on Build GRCh38
rs1031484095 Chr.7: 2360864-2360864 on Build GRCh38
rs763565370 Chr.7: 2360884-2360884 on Build GRCh38
rs777436544 Chr.7: 2360801-2360801 on Build GRCh38
rs934285697 Chr.7: 2360898-2360898 on Build GRCh38
rs368296808 Chr.7: 2360910-2360910 on Build GRCh38
rs770552909 Chr.7: 2360760-2360760 on Build GRCh38
rs776147143 Chr.7: 2360764-2360764 on Build GRCh38

View Full Product Details