Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49632210-49632449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTWD1
Entrez Gene ID
56986
Gene Name
DTW domain containing 1
Gene Aliases
MDS009
UniGene
Hs.127432
Gene Chromosome Location
Chr.15: 49620980-49654448 on Build GRCh38
Transcripts
NM_001144955.1 NM_020234.5 XM_011521815.1 XM_017022419.1 XM_017022420.1 XM_017022421.1 XM_017022422.1 XM_017022423.1 XM_017022424.1 XM_017022425.1 XM_017022426.1 XM_017022427.1 XM_017022428.1 XM_017022429.1 XM_017022430.1 XM_017022431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2216337 c.368C>T p.T123M Chr.15: 49632262-49632262 on Build GRCh38
4055410 c.322G>T p.A108S Chr.15: 49632216-49632216 on Build GRCh38
7127194 c.350A>C p.E117A Chr.15: 49632244-49632244 on Build GRCh38
6436966 c.382C>A p.P128T Chr.15: 49632276-49632276 on Build GRCh38
4517721 c.402_403CC>TT p.H135Y Chr.15: 49632296-49632297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754756313 Chr.15: 49632262-49632262 on Build GRCh38
rs201902941 Chr.15: 49632330-49632330 on Build GRCh38
rs777031190 Chr.15: 49632299-49632299 on Build GRCh38
rs1034584847 Chr.15: 49632366-49632366 on Build GRCh38
rs754630319 Chr.15: 49632337-49632337 on Build GRCh38
rs750934986 Chr.15: 49632325-49632325 on Build GRCh38
rs374894413 Chr.15: 49632328-49632328 on Build GRCh38
rs534291137 Chr.15: 49632362-49632362 on Build GRCh38
rs772056829 Chr.15: 49632287-49632287 on Build GRCh38
rs533175402 Chr.15: 49632315-49632315 on Build GRCh38
rs888864283 Chr.15: 49632404-49632404 on Build GRCh38
rs566810969 Chr.15: 49632358-49632358 on Build GRCh38
rs555625731 Chr.15: 49632363-49632363 on Build GRCh38
rs1010581197 Chr.15: 49632302-49632302 on Build GRCh38
rs769032945 Chr.15: 49632298-49632298 on Build GRCh38
rs751186028 Chr.15: 49632333-49632333 on Build GRCh38
rs146894806 Chr.15: 49632243-49632243 on Build GRCh38
rs769908340 Chr.15: 49632268-49632268 on Build GRCh38
rs371850152 Chr.15: 49632269-49632269 on Build GRCh38
rs758041917 Chr.15: 49632254-49632254 on Build GRCh38
rs1011780265 Chr.15: 49632405-49632405 on Build GRCh38
rs1029192904 Chr.15: 49632394-49632394 on Build GRCh38
rs781117355 Chr.15: 49632340-49632340 on Build GRCh38
rs1056878224 Chr.15: 49632281-49632281 on Build GRCh38
rs374556661 Chr.15: 49632242-49632242 on Build GRCh38
rs550081886 Chr.15: 49632265-49632265 on Build GRCh38
rs775731909 Chr.15: 49632291-49632291 on Build GRCh38
rs200808346 Chr.15: 49632263-49632263 on Build GRCh38
rs745873020 Chr.15: 49632284-49632284 on Build GRCh38
rs780056041 Chr.15: 49632338-49632339 on Build GRCh38
rs751188609 Chr.15: 49632260-49632260 on Build GRCh38
rs539419389 Chr.15: 49632301-49632301 on Build GRCh38
rs779694535 Chr.15: 49632257-49632257 on Build GRCh38
rs1041610017 Chr.15: 49632313-49632313 on Build GRCh38
rs999829005 Chr.15: 49632359-49632359 on Build GRCh38
rs760836248 Chr.15: 49632296-49632296 on Build GRCh38

View Full Product Details