Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8122940-8123177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES7
Entrez Gene ID
84667
Gene Name
hes family bHLH transcription factor 7
Gene Aliases
SCDO4, bHLHb37
UniGene
Hs.434828
Gene Chromosome Location
Chr.17: 8120590-8126032 on Build GRCh38
Transcripts
NM_001165967.1 NM_032580.3 XM_011524041.2 XM_017025232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382540 c.100G>T p.E34* Chr.17: 8123069-8123069 on Build GRCh38
4070893 c.93C>G p.S31R Chr.17: 8123076-8123076 on Build GRCh38
5387713 c.96G>A p.L32L Chr.17: 8123073-8123073 on Build GRCh38
6322227 c.84C>G p.I28M Chr.17: 8123085-8123085 on Build GRCh38
3388326 c.70C>A p.R24S Chr.17: 8123099-8123099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765170412 Chr.17: 8123101-8123101 on Build GRCh38
rs774790059 Chr.17: 8123068-8123068 on Build GRCh38
rs139906985 Chr.17: 8122961-8122961 on Build GRCh38
rs368725204 Chr.17: 8123013-8123013 on Build GRCh38
rs371748968 Chr.17: 8122986-8122986 on Build GRCh38
rs750181156 Chr.17: 8123110-8123110 on Build GRCh38
rs778047084 Chr.17: 8123050-8123050 on Build GRCh38
rs761667617 Chr.17: 8123099-8123099 on Build GRCh38
rs886043538 Chr.17: 8123022-8123022 on Build GRCh38
rs775539610 Chr.17: 8123133-8123133 on Build GRCh38
rs376311641 Chr.17: 8123028-8123028 on Build GRCh38
rs372627919 Chr.17: 8123090-8123090 on Build GRCh38
rs759833400 Chr.17: 8123076-8123076 on Build GRCh38
rs956534860 Chr.17: 8123073-8123073 on Build GRCh38
rs919342333 Chr.17: 8123041-8123041 on Build GRCh38
rs752195493 Chr.17: 8122992-8122992 on Build GRCh38
rs777996899 Chr.17: 8123140-8123140 on Build GRCh38
rs552373254 Chr.17: 8123003-8123003 on Build GRCh38
rs753408923 Chr.17: 8123094-8123094 on Build GRCh38
rs912479253 Chr.17: 8123138-8123138 on Build GRCh38
rs954644488 Chr.17: 8122968-8122968 on Build GRCh38
rs758157463 Chr.17: 8123117-8123117 on Build GRCh38
rs781290618 Chr.17: 8123039-8123039 on Build GRCh38
rs763576909 Chr.17: 8123091-8123091 on Build GRCh38
rs756416284 Chr.17: 8123136-8123136 on Build GRCh38
rs987042061 Chr.17: 8123133-8123133 on Build GRCh38
rs376049573 Chr.17: 8123008-8123008 on Build GRCh38
rs749800673 Chr.17: 8123055-8123055 on Build GRCh38
rs1021612889 Chr.17: 8122975-8122975 on Build GRCh38
rs986116992 Chr.17: 8122971-8122971 on Build GRCh38
rs771978237 Chr.17: 8123126-8123127 on Build GRCh38
rs771634586 Chr.17: 8123058-8123058 on Build GRCh38
rs1011210823 Chr.17: 8123111-8123111 on Build GRCh38
rs766234398 Chr.17: 8123130-8123130 on Build GRCh38
rs61731639 Chr.17: 8123046-8123046 on Build GRCh38
rs750219149 Chr.17: 8123038-8123038 on Build GRCh38
rs368880766 Chr.17: 8123081-8123081 on Build GRCh38
rs113994160 Chr.17: 8123096-8123096 on Build GRCh38
rs371599866 Chr.17: 8123059-8123059 on Build GRCh38

View Full Product Details