Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150466947-150467214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP8
Entrez Gene ID
155038
Gene Name
GTPase, IMAP family member 8
Gene Aliases
IAN-9, IAN6, IAN9, IANT
UniGene
Hs.647121
Gene Chromosome Location
Chr.7: 150450630-150479395 on Build GRCh38
Transcripts
NM_175571.3 XM_005249950.4 XM_005249951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924874 c.481T>A p.Y161N Chr.7: 150467179-150467179 on Build GRCh38
4155654 c.427T>A p.L143M Chr.7: 150467125-150467125 on Build GRCh38
3635932 c.274C>T p.P92S Chr.7: 150466972-150466972 on Build GRCh38
485037 c.404G>A p.R135Q Chr.7: 150467102-150467102 on Build GRCh38
1699555 c.412G>A p.D138N Chr.7: 150467110-150467110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758062268 Chr.7: 150467025-150467025 on Build GRCh38
rs148301701 Chr.7: 150467146-150467146 on Build GRCh38
rs746840063 Chr.7: 150467040-150467040 on Build GRCh38
rs779998996 Chr.7: 150467156-150467156 on Build GRCh38
rs909925348 Chr.7: 150466994-150466994 on Build GRCh38
rs763137255 Chr.7: 150466986-150466986 on Build GRCh38
rs761170315 Chr.7: 150467102-150467102 on Build GRCh38
rs750836899 Chr.7: 150467003-150467003 on Build GRCh38
rs745785913 Chr.7: 150467045-150467045 on Build GRCh38
rs780849961 Chr.7: 150467141-150467141 on Build GRCh38
rs145682936 Chr.7: 150467007-150467007 on Build GRCh38
rs145618535 Chr.7: 150467180-150467180 on Build GRCh38
rs376916445 Chr.7: 150467029-150467029 on Build GRCh38
rs752172592 Chr.7: 150467130-150467130 on Build GRCh38
rs149849472 Chr.7: 150467023-150467023 on Build GRCh38
rs765719798 Chr.7: 150467124-150467124 on Build GRCh38
rs772254030 Chr.7: 150467169-150467169 on Build GRCh38
rs757868006 Chr.7: 150467131-150467131 on Build GRCh38
rs983706109 Chr.7: 150467046-150467046 on Build GRCh38
rs774306095 Chr.7: 150467072-150467072 on Build GRCh38
rs777445544 Chr.7: 150467132-150467132 on Build GRCh38
rs760973666 Chr.7: 150467188-150467188 on Build GRCh38
rs769638956 Chr.7: 150467150-150467150 on Build GRCh38
rs757105707 Chr.7: 150467140-150467140 on Build GRCh38
rs143212982 Chr.7: 150467101-150467101 on Build GRCh38
rs866044980 Chr.7: 150467065-150467065 on Build GRCh38
rs754574138 Chr.7: 150467017-150467017 on Build GRCh38
rs746795576 Chr.7: 150467143-150467144 on Build GRCh38
rs930093322 Chr.7: 150466968-150466968 on Build GRCh38
rs149007858 Chr.7: 150467157-150467157 on Build GRCh38
rs76135975 Chr.7: 150467021-150467021 on Build GRCh38
rs754436007 Chr.7: 150467110-150467110 on Build GRCh38
rs766836722 Chr.7: 150467105-150467105 on Build GRCh38
rs759966278 Chr.7: 150467113-150467113 on Build GRCh38
rs1012337307 Chr.7: 150467128-150467128 on Build GRCh38
rs767787407 Chr.7: 150467000-150467000 on Build GRCh38
rs61748320 Chr.7: 150467067-150467067 on Build GRCh38
rs754277049 Chr.7: 150467012-150467012 on Build GRCh38
rs903724377 Chr.7: 150466972-150466972 on Build GRCh38
rs761942193 Chr.7: 150466997-150466997 on Build GRCh38
rs764062527 Chr.7: 150466992-150466992 on Build GRCh38
rs767618404 Chr.7: 150467137-150467137 on Build GRCh38
rs770684528 Chr.7: 150467043-150467043 on Build GRCh38
rs767031054 Chr.7: 150467008-150467008 on Build GRCh38
rs777702188 Chr.7: 150467134-150467134 on Build GRCh38
rs959171508 Chr.7: 150466998-150466998 on Build GRCh38
rs987842228 Chr.7: 150467166-150467166 on Build GRCh38

View Full Product Details