Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109858562-109858835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTP
Entrez Gene ID
51228
Gene Name
glycolipid transfer protein
Gene Aliases
-
UniGene
Hs.381256
Gene Chromosome Location
Chr.12: 109850943-109880488 on Build GRCh38
Transcripts
NM_016433.3 XM_011538434.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7105813 c.135C>G p.I45M Chr.12: 109858710-109858710 on Build GRCh38
6671840 c.150C>T p.S50S Chr.12: 109858695-109858695 on Build GRCh38
3935731 c.115T>C p.S39P Chr.12: 109858730-109858730 on Build GRCh38
1946722 c.151G>A p.G51S Chr.12: 109858694-109858694 on Build GRCh38
1946723 c.128C>T p.T43I Chr.12: 109858717-109858717 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs577886900 Chr.12: 109858597-109858597 on Build GRCh38
rs200730113 Chr.12: 109858668-109858668 on Build GRCh38
rs922070535 Chr.12: 109858606-109858606 on Build GRCh38
rs778721506 Chr.12: 109858705-109858705 on Build GRCh38
rs769641115 Chr.12: 109858803-109858803 on Build GRCh38
rs760078865 Chr.12: 109858741-109858741 on Build GRCh38
rs760619246 Chr.12: 109858666-109858666 on Build GRCh38
rs565458061 Chr.12: 109858787-109858787 on Build GRCh38
rs548087713 Chr.12: 109858813-109858813 on Build GRCh38
rs771407800 Chr.12: 109858701-109858701 on Build GRCh38
rs751786915 Chr.12: 109858774-109858774 on Build GRCh38
rs749710788 Chr.12: 109858698-109858698 on Build GRCh38
rs374151725 Chr.12: 109858678-109858678 on Build GRCh38
rs757129858 Chr.12: 109858724-109858724 on Build GRCh38
rs764958678 Chr.12: 109858767-109858767 on Build GRCh38
rs759717540 Chr.12: 109858775-109858775 on Build GRCh38
rs753712209 Chr.12: 109858669-109858669 on Build GRCh38
rs531425150 Chr.12: 109858740-109858740 on Build GRCh38
rs911716832 Chr.12: 109858682-109858682 on Build GRCh38
rs749286491 Chr.12: 109858686-109858686 on Build GRCh38
rs186646781 Chr.12: 109858662-109858662 on Build GRCh38
rs761657427 Chr.12: 109858762-109858762 on Build GRCh38
rs745919609 Chr.12: 109858645-109858645 on Build GRCh38
rs745444476 Chr.12: 109858707-109858707 on Build GRCh38
rs140183607 Chr.12: 109858695-109858695 on Build GRCh38
rs776542083 Chr.12: 109858755-109858755 on Build GRCh38
rs191494455 Chr.12: 109858784-109858784 on Build GRCh38
rs755066428 Chr.12: 109858683-109858683 on Build GRCh38
rs766992810 Chr.12: 109858658-109858658 on Build GRCh38
rs375880158 Chr.12: 109858802-109858802 on Build GRCh38
rs371042709 Chr.12: 109858644-109858644 on Build GRCh38
rs751772277 Chr.12: 109858681-109858681 on Build GRCh38
rs182929112 Chr.12: 109858791-109858791 on Build GRCh38
rs145689737 Chr.12: 109858712-109858712 on Build GRCh38
rs561014042 Chr.12: 109858721-109858721 on Build GRCh38
rs756596529 Chr.12: 109858694-109858694 on Build GRCh38
rs545309667 Chr.12: 109858646-109858646 on Build GRCh38
rs953601244 Chr.12: 109858609-109858609 on Build GRCh38
rs768675182 Chr.12: 109858754-109858754 on Build GRCh38
rs757011761 Chr.12: 109858674-109858674 on Build GRCh38
rs773587168 Chr.12: 109858635-109858635 on Build GRCh38
rs753176475 Chr.12: 109858687-109858687 on Build GRCh38

View Full Product Details