Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154350026-154350265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
UniGene
Hs.195464
Gene Chromosome Location
Chr.X: 154348529-154374638 on Build GRCh38
Transcripts
NM_001110556.1 NM_001456.3 XM_011531127.1 XM_011531128.1 XM_011531129.1 XM_011531130.1 XM_011531131.1

SNP Detail

SNP ID Chromosome Location
rs781903616 Chr.X: 154350214-154350214 on Build GRCh38
rs782289803 Chr.X: 154350105-154350105 on Build GRCh38
rs782430980 Chr.X: 154350234-154350234 on Build GRCh38
rs782267549 Chr.X: 154350243-154350243 on Build GRCh38
rs782541746 Chr.X: 154350197-154350197 on Build GRCh38
rs934536261 Chr.X: 154350188-154350188 on Build GRCh38
rs781967286 Chr.X: 154350225-154350225 on Build GRCh38
rs1016418093 Chr.X: 154350090-154350090 on Build GRCh38
rs782750091 Chr.X: 154350059-154350059 on Build GRCh38
rs781906091 Chr.X: 154350155-154350155 on Build GRCh38
rs368038166 Chr.X: 154350181-154350181 on Build GRCh38
rs202152952 Chr.X: 154350159-154350159 on Build GRCh38
rs782645927 Chr.X: 154350190-154350190 on Build GRCh38
rs781857682 Chr.X: 154350089-154350089 on Build GRCh38
rs12008807 Chr.X: 154350140-154350140 on Build GRCh38
rs782345328 Chr.X: 154350210-154350210 on Build GRCh38
rs782447267 Chr.X: 154350158-154350158 on Build GRCh38
rs782129907 Chr.X: 154350143-154350143 on Build GRCh38
rs782014331 Chr.X: 154350098-154350098 on Build GRCh38
rs782579933 Chr.X: 154350198-154350198 on Build GRCh38
rs782393028 Chr.X: 154350241-154350241 on Build GRCh38
rs878854463 Chr.X: 154350169-154350169 on Build GRCh38
rs201168500 Chr.X: 154350142-154350142 on Build GRCh38
rs727503930 Chr.X: 154350192-154350192 on Build GRCh38
rs375751609 Chr.X: 154350137-154350137 on Build GRCh38
rs371563857 Chr.X: 154350207-154350207 on Build GRCh38
rs200198847 Chr.X: 154350097-154350097 on Build GRCh38
rs886044822 Chr.X: 154350119-154350119 on Build GRCh38
rs781831266 Chr.X: 154350180-154350180 on Build GRCh38
rs782308324 Chr.X: 154350109-154350109 on Build GRCh38
rs782811927 Chr.X: 154350154-154350154 on Build GRCh38

View Full Product Details