Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147908190-147908415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA8
Entrez Gene ID
2703
Gene Name
gap junction protein alpha 8
Gene Aliases
CAE, CAE1, CTRCT1, CX50, CZP1, MP70
UniGene
Hs.632441
Gene Chromosome Location
Chr.1: 147902820-147915287 on Build GRCh38
Transcripts
NM_005267.4 XM_011509416.1 XM_011509417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1580944 c.317G>A p.R106H Chr.1: 147908272-147908272 on Build GRCh38
374932 c.294C>G p.H98Q Chr.1: 147908249-147908249 on Build GRCh38
5377529 c.435G>A p.G145G Chr.1: 147908390-147908390 on Build GRCh38
1579696 c.326G>A p.R109H Chr.1: 147908281-147908281 on Build GRCh38
4443155 c.353C>T p.A118V Chr.1: 147908308-147908308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782093190 Chr.1: 147908267-147908267 on Build GRCh38
rs782245059 Chr.1: 147908360-147908360 on Build GRCh38
rs782398614 Chr.1: 147908251-147908251 on Build GRCh38
rs781915687 Chr.1: 147908283-147908283 on Build GRCh38
rs145974798 Chr.1: 147908394-147908394 on Build GRCh38
rs200408514 Chr.1: 147908243-147908243 on Build GRCh38
rs926855257 Chr.1: 147908233-147908233 on Build GRCh38
rs782044825 Chr.1: 147908253-147908253 on Build GRCh38
rs782214202 Chr.1: 147908310-147908310 on Build GRCh38
rs886045254 Chr.1: 147908255-147908255 on Build GRCh38
rs886045253 Chr.1: 147908240-147908240 on Build GRCh38
rs782348605 Chr.1: 147908232-147908232 on Build GRCh38
rs782367791 Chr.1: 147908396-147908396 on Build GRCh38
rs200110674 Chr.1: 147908392-147908392 on Build GRCh38
rs782199122 Chr.1: 147908218-147908218 on Build GRCh38
rs372885882 Chr.1: 147908288-147908288 on Build GRCh38
rs781949456 Chr.1: 147908257-147908257 on Build GRCh38
rs782663112 Chr.1: 147908298-147908298 on Build GRCh38
rs1042509090 Chr.1: 147908302-147908302 on Build GRCh38
rs782766869 Chr.1: 147908350-147908350 on Build GRCh38
rs782185896 Chr.1: 147908355-147908355 on Build GRCh38
rs782558352 Chr.1: 147908354-147908354 on Build GRCh38
rs782569623 Chr.1: 147908214-147908214 on Build GRCh38
rs371709557 Chr.1: 147908347-147908347 on Build GRCh38
rs782457351 Chr.1: 147908281-147908281 on Build GRCh38
rs782313023 Chr.1: 147908381-147908381 on Build GRCh38
rs80358200 Chr.1: 147908217-147908217 on Build GRCh38
rs782125496 Chr.1: 147908321-147908321 on Build GRCh38
rs781792753 Chr.1: 147908280-147908280 on Build GRCh38
rs782714532 Chr.1: 147908336-147908336 on Build GRCh38
rs782294604 Chr.1: 147908380-147908380 on Build GRCh38
rs782119026 Chr.1: 147908249-147908249 on Build GRCh38
rs782460854 Chr.1: 147908357-147908357 on Build GRCh38
rs782232746 Chr.1: 147908294-147908294 on Build GRCh38
rs782048169 Chr.1: 147908328-147908328 on Build GRCh38
rs200587705 Chr.1: 147908311-147908311 on Build GRCh38
rs376298781 Chr.1: 147908358-147908358 on Build GRCh38
rs781981299 Chr.1: 147908312-147908312 on Build GRCh38
rs142426709 Chr.1: 147908252-147908252 on Build GRCh38
rs782036781 Chr.1: 147908327-147908327 on Build GRCh38
rs782623142 Chr.1: 147908276-147908276 on Build GRCh38
rs782014995 Chr.1: 147908376-147908376 on Build GRCh38
rs142415337 Chr.1: 147908340-147908340 on Build GRCh38
rs367754368 Chr.1: 147908307-147908307 on Build GRCh38
rs781873537 Chr.1: 147908353-147908353 on Build GRCh38
rs782741157 Chr.1: 147908222-147908222 on Build GRCh38
rs781901664 Chr.1: 147908374-147908374 on Build GRCh38
rs917690151 Chr.1: 147908272-147908272 on Build GRCh38
rs374557851 Chr.1: 147908295-147908295 on Build GRCh38
rs900423232 Chr.1: 147908326-147908326 on Build GRCh38
rs142026480 Chr.1: 147908322-147908322 on Build GRCh38
rs886242039 Chr.1: 147908364-147908364 on Build GRCh38

View Full Product Details