Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 92655117-92655388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM174B
Entrez Gene ID
400451
Gene Name
family with sequence similarity 174 member B
Gene Aliases
-
UniGene
Hs.27373
Gene Chromosome Location
Chr.15: 92617447-92734219 on Build GRCh38
Transcripts
NM_207446.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4578631 c.320C>T p.A107V Chr.15: 92655340-92655340 on Build GRCh38
6421343 c.289G>A p.V97M Chr.15: 92655371-92655371 on Build GRCh38
966686 c.318C>A p.I106I Chr.15: 92655342-92655342 on Build GRCh38
4925300 c.309C>A p.T103T Chr.15: 92655351-92655351 on Build GRCh38
6298180 c.293C>A p.A98E Chr.15: 92655367-92655367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768378186 Chr.15: 92655308-92655308 on Build GRCh38
rs751937647 Chr.15: 92655360-92655360 on Build GRCh38
rs1036146357 Chr.15: 92655190-92655190 on Build GRCh38
rs759577915 Chr.15: 92655348-92655348 on Build GRCh38
rs1006389734 Chr.15: 92655157-92655157 on Build GRCh38
rs367642570 Chr.15: 92655298-92655298 on Build GRCh38
rs887946996 Chr.15: 92655147-92655147 on Build GRCh38
rs778308570 Chr.15: 92655291-92655291 on Build GRCh38
rs984644964 Chr.15: 92655164-92655164 on Build GRCh38
rs909114248 Chr.15: 92655177-92655177 on Build GRCh38
rs578000700 Chr.15: 92655361-92655361 on Build GRCh38
rs570409026 Chr.15: 92655238-92655238 on Build GRCh38
rs761803045 Chr.15: 92655323-92655323 on Build GRCh38
rs775242366 Chr.15: 92655272-92655272 on Build GRCh38
rs984082456 Chr.15: 92655280-92655280 on Build GRCh38
rs761277680 Chr.15: 92655286-92655287 on Build GRCh38
rs953042507 Chr.15: 92655219-92655219 on Build GRCh38
rs774132861 Chr.15: 92655318-92655318 on Build GRCh38
rs547102260 Chr.15: 92655226-92655226 on Build GRCh38
rs949114438 Chr.15: 92655234-92655234 on Build GRCh38
rs372531244 Chr.15: 92655367-92655367 on Build GRCh38
rs763905775 Chr.15: 92655275-92655275 on Build GRCh38
rs138235581 Chr.15: 92655158-92655158 on Build GRCh38
rs867772863 Chr.15: 92655202-92655202 on Build GRCh38
rs748869488 Chr.15: 92655371-92655371 on Build GRCh38
rs1053443694 Chr.15: 92655359-92655359 on Build GRCh38
rs746294046 Chr.15: 92655271-92655278 on Build GRCh38
rs555935163 Chr.15: 92655248-92655248 on Build GRCh38
rs375712759 Chr.15: 92655292-92655297 on Build GRCh38
rs772407210 Chr.15: 92655283-92655298 on Build GRCh38
rs776019584 Chr.15: 92655283-92655284 on Build GRCh38
rs371564462 Chr.15: 92655352-92655352 on Build GRCh38
rs765443275 Chr.15: 92655349-92655349 on Build GRCh38
rs758798317 Chr.15: 92655286-92655286 on Build GRCh38
rs892722662 Chr.15: 92655254-92655254 on Build GRCh38
rs77720151 Chr.15: 92655332-92655332 on Build GRCh38
rs114877871 Chr.15: 92655304-92655304 on Build GRCh38
rs924555371 Chr.15: 92655252-92655252 on Build GRCh38
rs960248465 Chr.15: 92655265-92655265 on Build GRCh38
rs965578315 Chr.15: 92655271-92655271 on Build GRCh38
rs539420103 Chr.15: 92655240-92655240 on Build GRCh38
rs555681253 Chr.15: 92655289-92655289 on Build GRCh38
rs747493526 Chr.15: 92655292-92655292 on Build GRCh38
rs1000519094 Chr.15: 92655242-92655242 on Build GRCh38
rs200954392 Chr.15: 92655277-92655277 on Build GRCh38
rs770296182 Chr.15: 92655215-92655216 on Build GRCh38
rs778216819 Chr.15: 92655354-92655354 on Build GRCh38
rs766447323 Chr.15: 92655183-92655183 on Build GRCh38
rs762776866 Chr.15: 92655273-92655273 on Build GRCh38
rs764865951 Chr.15: 92655266-92655266 on Build GRCh38
rs1009859182 Chr.15: 92655257-92655257 on Build GRCh38
rs368855850 Chr.15: 92655363-92655363 on Build GRCh38
rs755211070 Chr.15: 92655279-92655279 on Build GRCh38
rs528630729 Chr.15: 92655180-92655180 on Build GRCh38
rs369743222 Chr.15: 92655350-92655350 on Build GRCh38
rs371711774 Chr.15: 92655303-92655303 on Build GRCh38
rs766412304 Chr.15: 92655278-92655278 on Build GRCh38
rs746416191 Chr.15: 92655362-92655362 on Build GRCh38
rs371887853 Chr.15: 92655342-92655342 on Build GRCh38
rs771349289 Chr.15: 92655296-92655296 on Build GRCh38
rs1030426643 Chr.15: 92655198-92655198 on Build GRCh38
rs997212984 Chr.15: 92655184-92655184 on Build GRCh38
rs780750173 Chr.15: 92655297-92655297 on Build GRCh38
rs503112 Chr.15: 92655283-92655283 on Build GRCh38

View Full Product Details