Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206966941-206967214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCAMR
Entrez Gene ID
83953
Gene Name
Fc fragment of IgA and IgM receptor
Gene Aliases
CD351, FCA/MR, FKSG87
UniGene
Hs.145519
Gene Chromosome Location
Chr.1: 206957955-206970657 on Build GRCh38
Transcripts
NM_001122979.2 NM_001170631.1 NM_032029.4 XM_011510051.2 XM_011510052.2 XM_017002442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
903059 c.138G>A p.M46I Chr.1: 206967083-206967083 on Build GRCh38
6350544 c.126G>A p.A42A Chr.1: 206967095-206967095 on Build GRCh38
146750 c.126G>A p.A42A Chr.1: 206967095-206967095 on Build GRCh38
1583976 c.138G>A p.M46I Chr.1: 206967083-206967083 on Build GRCh38
6571737 c.169+1G>T p.? Chr.1: 206967051-206967051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909928808 Chr.1: 206967139-206967139 on Build GRCh38
rs959969766 Chr.1: 206967084-206967084 on Build GRCh38
rs999028285 Chr.1: 206967116-206967117 on Build GRCh38
rs763772325 Chr.1: 206967138-206967138 on Build GRCh38
rs200414607 Chr.1: 206967104-206967104 on Build GRCh38
rs753100249 Chr.1: 206967127-206967127 on Build GRCh38
rs977245248 Chr.1: 206967049-206967049 on Build GRCh38
rs1021304018 Chr.1: 206967022-206967022 on Build GRCh38
rs751221229 Chr.1: 206967147-206967147 on Build GRCh38
rs780022708 Chr.1: 206967161-206967161 on Build GRCh38
rs765856357 Chr.1: 206967123-206967123 on Build GRCh38
rs907112655 Chr.1: 206966970-206966970 on Build GRCh38
rs1028050614 Chr.1: 206966992-206966992 on Build GRCh38
rs749938693 Chr.1: 206967156-206967156 on Build GRCh38
rs199746455 Chr.1: 206967016-206967016 on Build GRCh38
rs758223260 Chr.1: 206967125-206967126 on Build GRCh38
rs990577722 Chr.1: 206967174-206967174 on Build GRCh38
rs779785248 Chr.1: 206967143-206967144 on Build GRCh38
rs939962089 Chr.1: 206966971-206966971 on Build GRCh38
rs373515731 Chr.1: 206967101-206967101 on Build GRCh38
rs995594312 Chr.1: 206966982-206966982 on Build GRCh38
rs143261323 Chr.1: 206967036-206967036 on Build GRCh38
rs776868901 Chr.1: 206967113-206967113 on Build GRCh38
rs898130117 Chr.1: 206966977-206966977 on Build GRCh38
rs376694888 Chr.1: 206967080-206967080 on Build GRCh38
rs1037194826 Chr.1: 206967157-206967157 on Build GRCh38
rs773370049 Chr.1: 206967096-206967096 on Build GRCh38
rs778621841 Chr.1: 206966974-206966974 on Build GRCh38
rs760057693 Chr.1: 206967121-206967121 on Build GRCh38
rs769754760 Chr.1: 206967062-206967062 on Build GRCh38
rs866071622 Chr.1: 206967144-206967144 on Build GRCh38
rs749077293 Chr.1: 206967163-206967163 on Build GRCh38
rs746528416 Chr.1: 206967003-206967003 on Build GRCh38
rs199884709 Chr.1: 206967102-206967102 on Build GRCh38
rs755988122 Chr.1: 206967159-206967159 on Build GRCh38
rs1036955704 Chr.1: 206966972-206966972 on Build GRCh38
rs780642279 Chr.1: 206967154-206967154 on Build GRCh38
rs369666643 Chr.1: 206967128-206967128 on Build GRCh38
rs745757062 Chr.1: 206967053-206967053 on Build GRCh38
rs11119933 Chr.1: 206967095-206967095 on Build GRCh38
rs1031954183 Chr.1: 206967067-206967067 on Build GRCh38

View Full Product Details