Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 76502730-76502989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT1
Entrez Gene ID
2650
Gene Name
glucosaminyl (N-acetyl) transferase 1, core 2
Gene Aliases
C2GNT, C2GNT-L, C2GNT1, G6NT, NACGT2, NAGCT2
UniGene
Hs.521568
Gene Chromosome Location
Chr.9: 76441666-76507416 on Build GRCh38
Transcripts
NM_001097633.1 NM_001097634.1 NM_001097635.1 NM_001097636.1 NM_001490.4 XM_017014620.1 XM_017014621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6184146 c.385G>T p.V129F Chr.9: 76502766-76502766 on Build GRCh38
6193910 c.608T>G p.L203R Chr.9: 76502989-76502989 on Build GRCh38
754212 c.469A>G p.K157E Chr.9: 76502850-76502850 on Build GRCh38
6116324 c.591C>G p.L197L Chr.9: 76502972-76502972 on Build GRCh38
7369337 c.473C>G p.S158C Chr.9: 76502854-76502854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746423733 Chr.9: 76502945-76502945 on Build GRCh38
rs749840186 Chr.9: 76502753-76502753 on Build GRCh38
rs759615039 Chr.9: 76502769-76502769 on Build GRCh38
rs150529677 Chr.9: 76502787-76502787 on Build GRCh38
rs778230598 Chr.9: 76502844-76502844 on Build GRCh38
rs750290107 Chr.9: 76502798-76502798 on Build GRCh38
rs780811950 Chr.9: 76502834-76502834 on Build GRCh38
rs975587504 Chr.9: 76502761-76502761 on Build GRCh38
rs754231347 Chr.9: 76502938-76502938 on Build GRCh38
rs758387360 Chr.9: 76502806-76502806 on Build GRCh38
rs774224861 Chr.9: 76502852-76502852 on Build GRCh38
rs770875325 Chr.9: 76502760-76502760 on Build GRCh38
rs71509884 Chr.9: 76502963-76502963 on Build GRCh38
rs775826802 Chr.9: 76502856-76502856 on Build GRCh38
rs149948942 Chr.9: 76502955-76502955 on Build GRCh38
rs769007670 Chr.9: 76502967-76502967 on Build GRCh38
rs2282683 Chr.9: 76502835-76502835 on Build GRCh38
rs757625842 Chr.9: 76502941-76502941 on Build GRCh38
rs754550197 Chr.9: 76502815-76502815 on Build GRCh38
rs771478745 Chr.9: 76502849-76502849 on Build GRCh38
rs756437872 Chr.9: 76502842-76502842 on Build GRCh38
rs755785501 Chr.9: 76502931-76502931 on Build GRCh38
rs758040032 Chr.9: 76502812-76502812 on Build GRCh38
rs762732983 Chr.9: 76502901-76502901 on Build GRCh38
rs766336364 Chr.9: 76502920-76502920 on Build GRCh38
rs570491038 Chr.9: 76502922-76502922 on Build GRCh38
rs773075818 Chr.9: 76502889-76502889 on Build GRCh38
rs141640466 Chr.9: 76502779-76502779 on Build GRCh38
rs767852982 Chr.9: 76502777-76502777 on Build GRCh38
rs1043099556 Chr.9: 76502828-76502828 on Build GRCh38
rs777941377 Chr.9: 76502933-76502933 on Build GRCh38
rs140433187 Chr.9: 76502796-76502796 on Build GRCh38
rs139675992 Chr.9: 76502886-76502886 on Build GRCh38
rs761011045 Chr.9: 76502868-76502868 on Build GRCh38
rs763744548 Chr.9: 76502928-76502929 on Build GRCh38
rs11546569 Chr.9: 76502854-76502854 on Build GRCh38
rs749708931 Chr.9: 76502846-76502846 on Build GRCh38
rs774511006 Chr.9: 76502767-76502767 on Build GRCh38
rs546314870 Chr.9: 76502948-76502948 on Build GRCh38
rs772333502 Chr.9: 76502855-76502855 on Build GRCh38
rs779459124 Chr.9: 76502944-76502944 on Build GRCh38
rs759026554 Chr.9: 76502923-76502923 on Build GRCh38
rs539128019 Chr.9: 76502930-76502930 on Build GRCh38
rs373599348 Chr.9: 76502814-76502814 on Build GRCh38

View Full Product Details