Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99711988-99712261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRRS1
Entrez Gene ID
391059
Gene Name
ferric chelate reductase 1
Gene Aliases
SDFR2, SDR2
UniGene
Hs.454779
Gene Chromosome Location
Chr.1: 99708699-99766630 on Build GRCh38
Transcripts
NM_001013660.2 XM_005270861.3 XM_011541451.2 XM_011541452.1 XM_017001271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5991655 c.1480+5_1480+8delGAGA p.? Chr.1: 99712097-99712100 on Build GRCh38
6500138 c.1452_1480+7del36 p.? Chr.1: 99712098-99712133 on Build GRCh38
1472325 c.1469G>A p.R490K Chr.1: 99712116-99712116 on Build GRCh38
5531475 c.1444C>T p.H482Y Chr.1: 99712141-99712141 on Build GRCh38
7095115 c.1422-1G>A p.? Chr.1: 99712164-99712164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772425613 Chr.1: 99712141-99712141 on Build GRCh38
rs900935078 Chr.1: 99712219-99712219 on Build GRCh38
rs373272274 Chr.1: 99712079-99712079 on Build GRCh38
rs200663740 Chr.1: 99712153-99712153 on Build GRCh38
rs61740680 Chr.1: 99712143-99712143 on Build GRCh38
rs374061776 Chr.1: 99712161-99712161 on Build GRCh38
rs750591091 Chr.1: 99712068-99712068 on Build GRCh38
rs991335124 Chr.1: 99712019-99712019 on Build GRCh38
rs1048346731 Chr.1: 99712054-99712054 on Build GRCh38
rs374512701 Chr.1: 99712198-99712198 on Build GRCh38
rs372348291 Chr.1: 99712211-99712211 on Build GRCh38
rs945848484 Chr.1: 99712221-99712221 on Build GRCh38
rs116770015 Chr.1: 99712099-99712099 on Build GRCh38
rs148375757 Chr.1: 99712139-99712139 on Build GRCh38
rs1025128211 Chr.1: 99712085-99712085 on Build GRCh38
rs746066118 Chr.1: 99712228-99712228 on Build GRCh38
rs755091150 Chr.1: 99712111-99712111 on Build GRCh38
rs148755843 Chr.1: 99712087-99712087 on Build GRCh38
rs1033258301 Chr.1: 99712226-99712226 on Build GRCh38
rs776383435 Chr.1: 99712162-99712162 on Build GRCh38
rs41285732 Chr.1: 99712167-99712167 on Build GRCh38
rs779372849 Chr.1: 99712131-99712131 on Build GRCh38
rs746620454 Chr.1: 99712150-99712150 on Build GRCh38

View Full Product Details