Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 5430964-5431229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARS2
Entrez Gene ID
10667
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD14, FARS1, HSPC320, PheRS
UniGene
Hs.484547
Gene Chromosome Location
Chr.6: 5261001-5771592 on Build GRCh38
Transcripts
NM_001318872.1 NM_006567.4 XM_005248812.3 XM_006714966.2 XM_011514247.2 XM_011514248.2 XM_011514249.1 XM_011514251.2 XM_017010186.1 XM_017010187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884397 c.817C>T p.P273S Chr.6: 5431085-5431085 on Build GRCh38
3928551 c.888A>G p.Q296Q Chr.6: 5431156-5431156 on Build GRCh38
5870931 c.850G>A p.E284K Chr.6: 5431118-5431118 on Build GRCh38
6350352 c.773-9A>T p.? Chr.6: 5431032-5431032 on Build GRCh38
742491 c.848G>A p.G283E Chr.6: 5431116-5431116 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779613121 Chr.6: 5431000-5431000 on Build GRCh38
rs758012144 Chr.6: 5430994-5430994 on Build GRCh38
rs754646580 Chr.6: 5431184-5431184 on Build GRCh38
rs373299744 Chr.6: 5431202-5431202 on Build GRCh38
rs746201644 Chr.6: 5431043-5431043 on Build GRCh38
rs780671245 Chr.6: 5431016-5431016 on Build GRCh38
rs886256686 Chr.6: 5431150-5431150 on Build GRCh38
rs746533036 Chr.6: 5431011-5431011 on Build GRCh38
rs863223959 Chr.6: 5431167-5431167 on Build GRCh38
rs372301183 Chr.6: 5431141-5431141 on Build GRCh38
rs761915432 Chr.6: 5431145-5431145 on Build GRCh38
rs773257335 Chr.6: 5431151-5431151 on Build GRCh38
rs749799074 Chr.6: 5431019-5431019 on Build GRCh38
rs559278551 Chr.6: 5431203-5431203 on Build GRCh38
rs778616479 Chr.6: 5430992-5430992 on Build GRCh38
rs1004641643 Chr.6: 5431200-5431200 on Build GRCh38
rs576701962 Chr.6: 5431026-5431026 on Build GRCh38
rs761709212 Chr.6: 5431069-5431069 on Build GRCh38
rs575864272 Chr.6: 5431085-5431085 on Build GRCh38
rs758127974 Chr.6: 5431103-5431103 on Build GRCh38
rs765097510 Chr.6: 5431148-5431148 on Build GRCh38
rs764139130 Chr.6: 5431114-5431114 on Build GRCh38
rs764336106 Chr.6: 5431065-5431065 on Build GRCh38
rs779412783 Chr.6: 5431115-5431115 on Build GRCh38
rs746233249 Chr.6: 5431117-5431117 on Build GRCh38
rs747278433 Chr.6: 5431142-5431142 on Build GRCh38
rs375267240 Chr.6: 5431002-5431002 on Build GRCh38
rs911419325 Chr.6: 5431024-5431024 on Build GRCh38
rs766146229 Chr.6: 5431105-5431105 on Build GRCh38
rs762869568 Chr.6: 5431163-5431163 on Build GRCh38
rs368336020 Chr.6: 5431058-5431058 on Build GRCh38
rs201927340 Chr.6: 5431049-5431049 on Build GRCh38
rs780728495 Chr.6: 5431112-5431112 on Build GRCh38
rs750213806 Chr.6: 5430993-5430993 on Build GRCh38
rs751161767 Chr.6: 5431004-5431004 on Build GRCh38
rs758942536 Chr.6: 5431205-5431205 on Build GRCh38
rs994991110 Chr.6: 5431131-5431131 on Build GRCh38
rs201042275 Chr.6: 5431087-5431087 on Build GRCh38
rs767156316 Chr.6: 5431188-5431188 on Build GRCh38
rs1034582028 Chr.6: 5431175-5431175 on Build GRCh38
rs376496854 Chr.6: 5431204-5431204 on Build GRCh38
rs760967282 Chr.6: 5431051-5431051 on Build GRCh38
rs772624228 Chr.6: 5431120-5431120 on Build GRCh38
rs776745910 Chr.6: 5431068-5431068 on Build GRCh38
rs201412926 Chr.6: 5431181-5431181 on Build GRCh38
rs11243011 Chr.6: 5431107-5431107 on Build GRCh38
rs375858088 Chr.6: 5431172-5431172 on Build GRCh38
rs768208541 Chr.6: 5431012-5431012 on Build GRCh38
rs776599876 Chr.6: 5431144-5431144 on Build GRCh38
rs761097220 Chr.6: 5431060-5431060 on Build GRCh38
rs773225879 Chr.6: 5431188-5431188 on Build GRCh38
rs752150366 Chr.6: 5431195-5431195 on Build GRCh38
rs936794507 Chr.6: 5430997-5430997 on Build GRCh38
rs774599491 Chr.6: 5431035-5431035 on Build GRCh38
rs748157826 Chr.6: 5431003-5431003 on Build GRCh38
rs771446019 Chr.6: 5431027-5431027 on Build GRCh38

View Full Product Details