Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41584746-41584973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP4
Entrez Gene ID
116113
Gene Name
forkhead box P4
Gene Aliases
hFKHLA
UniGene
Hs.131436
Gene Chromosome Location
Chr.6: 41546426-41602384 on Build GRCh38
Transcripts
NM_001012426.1 NM_001012427.1 NM_138457.2 XM_006714991.3 XM_011514289.2 XM_011514290.2 XM_011514291.2 XM_011514292.2 XM_011514293.2 XM_017010233.1 XM_017010234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5486564 c.311C>T p.S104L Chr.6: 41584779-41584779 on Build GRCh38
6233816 c.385G>T p.A129S Chr.6: 41584853-41584853 on Build GRCh38
1634797 c.342C>A p.T114T Chr.6: 41584810-41584810 on Build GRCh38
6233815 c.385G>T p.A129S Chr.6: 41584853-41584853 on Build GRCh38
7448340 c.391C>T p.L131F Chr.6: 41584859-41584859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770809515 Chr.6: 41584780-41584780 on Build GRCh38
rs772926947 Chr.6: 41584769-41584769 on Build GRCh38
rs760639636 Chr.6: 41584775-41584775 on Build GRCh38
rs761157934 Chr.6: 41584917-41584917 on Build GRCh38
rs546916123 Chr.6: 41584935-41584935 on Build GRCh38
rs762723447 Chr.6: 41584908-41584908 on Build GRCh38
rs757873080 Chr.6: 41584763-41584763 on Build GRCh38
rs776612898 Chr.6: 41584789-41584789 on Build GRCh38
rs756710342 Chr.6: 41584813-41584813 on Build GRCh38
rs932384357 Chr.6: 41584874-41584874 on Build GRCh38
rs771910838 Chr.6: 41584766-41584766 on Build GRCh38
rs1010515550 Chr.6: 41584882-41584882 on Build GRCh38
rs922357095 Chr.6: 41584859-41584859 on Build GRCh38
rs780473990 Chr.6: 41584818-41584818 on Build GRCh38
rs779033846 Chr.6: 41584824-41584824 on Build GRCh38
rs893507983 Chr.6: 41584880-41584880 on Build GRCh38
rs1049864098 Chr.6: 41584876-41584876 on Build GRCh38
rs763789652 Chr.6: 41584913-41584913 on Build GRCh38
rs752352374 Chr.6: 41584795-41584795 on Build GRCh38
rs754356891 Chr.6: 41584926-41584926 on Build GRCh38
rs764800851 Chr.6: 41584794-41584794 on Build GRCh38
rs774999702 Chr.6: 41584881-41584881 on Build GRCh38
rs751233902 Chr.6: 41584914-41584914 on Build GRCh38
rs772244259 Chr.6: 41584832-41584832 on Build GRCh38
rs755075359 Chr.6: 41584823-41584823 on Build GRCh38
rs746858403 Chr.6: 41584842-41584842 on Build GRCh38
rs933300031 Chr.6: 41584855-41584855 on Build GRCh38
rs201819185 Chr.6: 41584883-41584883 on Build GRCh38
rs766757684 Chr.6: 41584918-41584918 on Build GRCh38
rs754335515 Chr.6: 41584819-41584819 on Build GRCh38
rs368554446 Chr.6: 41584932-41584932 on Build GRCh38
rs748380841 Chr.6: 41584826-41584826 on Build GRCh38
rs750775706 Chr.6: 41584812-41584812 on Build GRCh38
rs772707860 Chr.6: 41584837-41584857 on Build GRCh38
rs199720815 Chr.6: 41584808-41584808 on Build GRCh38
rs1041193874 Chr.6: 41584848-41584848 on Build GRCh38
rs745997904 Chr.6: 41584798-41584798 on Build GRCh38
rs781747834 Chr.6: 41584797-41584797 on Build GRCh38
rs776499316 Chr.6: 41584857-41584857 on Build GRCh38
rs770686504 Chr.6: 41584853-41584853 on Build GRCh38
rs371450432 Chr.6: 41584802-41584802 on Build GRCh38
rs777570267 Chr.6: 41584837-41584837 on Build GRCh38
rs759009187 Chr.6: 41584791-41584791 on Build GRCh38
rs369513950 Chr.6: 41584861-41584861 on Build GRCh38
rs769203082 Chr.6: 41584863-41584863 on Build GRCh38
rs755452339 Chr.6: 41584928-41584928 on Build GRCh38

View Full Product Details