Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89264629-89264864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP5
Entrez Gene ID
115362
Gene Name
guanylate binding protein 5
Gene Aliases
GBP-5
UniGene
Hs.513726
Gene Chromosome Location
Chr.1: 89258950-89272861 on Build GRCh38
Transcripts
NM_001134486.2 NM_052942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4458459 c.1086C>T p.A362A Chr.1: 89264749-89264749 on Build GRCh38
6489727 c.1068G>T p.R356S Chr.1: 89264767-89264767 on Build GRCh38
1320854 c.981A>G p.Q327Q Chr.1: 89264854-89264854 on Build GRCh38
5352545 c.1106delA p.N369fs*6 Chr.1: 89264729-89264729 on Build GRCh38
3419552 c.1128A>C p.Q376H Chr.1: 89264707-89264707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143883598 Chr.1: 89264835-89264835 on Build GRCh38
rs200985091 Chr.1: 89264715-89264715 on Build GRCh38
rs752651991 Chr.1: 89264736-89264736 on Build GRCh38
rs775336799 Chr.1: 89264840-89264840 on Build GRCh38
rs776355366 Chr.1: 89264712-89264712 on Build GRCh38
rs1020017140 Chr.1: 89264689-89264689 on Build GRCh38
rs770529670 Chr.1: 89264827-89264827 on Build GRCh38
rs769330639 Chr.1: 89264690-89264690 on Build GRCh38
rs781424147 Chr.1: 89264656-89264656 on Build GRCh38
rs113830951 Chr.1: 89264834-89264834 on Build GRCh38
rs773978617 Chr.1: 89264691-89264691 on Build GRCh38
rs767496198 Chr.1: 89264747-89264747 on Build GRCh38
rs779558415 Chr.1: 89264775-89264775 on Build GRCh38
rs771554115 Chr.1: 89264795-89264795 on Build GRCh38
rs759276087 Chr.1: 89264836-89264836 on Build GRCh38
rs34832463 Chr.1: 89264722-89264723 on Build GRCh38
rs200417227 Chr.1: 89264845-89264845 on Build GRCh38
rs368074195 Chr.1: 89264837-89264837 on Build GRCh38
rs371973500 Chr.1: 89264722-89264722 on Build GRCh38
rs202079327 Chr.1: 89264701-89264701 on Build GRCh38
rs750597229 Chr.1: 89264748-89264748 on Build GRCh38
rs368263405 Chr.1: 89264717-89264717 on Build GRCh38
rs772656612 Chr.1: 89264811-89264811 on Build GRCh38
rs746667851 Chr.1: 89264817-89264817 on Build GRCh38
rs749812638 Chr.1: 89264687-89264687 on Build GRCh38
rs149475068 Chr.1: 89264707-89264707 on Build GRCh38
rs746105643 Chr.1: 89264673-89264673 on Build GRCh38
rs772855977 Chr.1: 89264710-89264710 on Build GRCh38
rs779523887 Chr.1: 89264762-89264762 on Build GRCh38
rs267598756 Chr.1: 89264737-89264737 on Build GRCh38
rs780439624 Chr.1: 89264756-89264756 on Build GRCh38
rs766185031 Chr.1: 89264711-89264711 on Build GRCh38
rs756361837 Chr.1: 89264751-89264751 on Build GRCh38
rs749722724 Chr.1: 89264758-89264758 on Build GRCh38
rs200752135 Chr.1: 89264676-89264676 on Build GRCh38
rs374866943 Chr.1: 89264794-89264794 on Build GRCh38
rs755419137 Chr.1: 89264761-89264761 on Build GRCh38
rs756430600 Chr.1: 89264674-89264674 on Build GRCh38

View Full Product Details