Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69385800-69386049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A2
Entrez Gene ID
9413
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61, X123
UniGene
Hs.118003
Gene Chromosome Location
Chr.9: 69324572-69392461 on Build GRCh38
Transcripts
NM_001127608.1 NM_004816.3 XM_005252307.4 XM_017015324.1 XM_017015325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109718 c.824G>A p.S275N Chr.9: 69385915-69385915 on Build GRCh38
5923838 c.841C>T p.P281S Chr.9: 69385932-69385932 on Build GRCh38
6396911 c.723A>G p.I241M Chr.9: 69385814-69385814 on Build GRCh38
3930098 c.856+1G>T p.? Chr.9: 69385948-69385948 on Build GRCh38
5718389 c.775C>T p.P259S Chr.9: 69385866-69385866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758313886 Chr.9: 69385831-69385831 on Build GRCh38
rs374544658 Chr.9: 69385996-69385996 on Build GRCh38
rs751368006 Chr.9: 69385842-69385842 on Build GRCh38
rs374537449 Chr.9: 69385915-69385915 on Build GRCh38
rs748266947 Chr.9: 69385863-69385863 on Build GRCh38
rs201780959 Chr.9: 69385956-69385956 on Build GRCh38
rs150027118 Chr.9: 69385879-69385879 on Build GRCh38
rs780028606 Chr.9: 69385837-69385837 on Build GRCh38
rs1037483071 Chr.9: 69386024-69386024 on Build GRCh38
rs768289768 Chr.9: 69385855-69385855 on Build GRCh38
rs749975561 Chr.9: 69385825-69385825 on Build GRCh38
rs762520444 Chr.9: 69385921-69385921 on Build GRCh38
rs914647894 Chr.9: 69385911-69385911 on Build GRCh38
rs149290712 Chr.9: 69386012-69386012 on Build GRCh38
rs11138395 Chr.9: 69385824-69385824 on Build GRCh38
rs779060681 Chr.9: 69385973-69385973 on Build GRCh38
rs781052620 Chr.9: 69385856-69385856 on Build GRCh38
rs888825898 Chr.9: 69386009-69386009 on Build GRCh38
rs764918218 Chr.9: 69385914-69385914 on Build GRCh38
rs746389535 Chr.9: 69385905-69385905 on Build GRCh38
rs377626202 Chr.9: 69385954-69385954 on Build GRCh38
rs756378943 Chr.9: 69385967-69385967 on Build GRCh38
rs772675960 Chr.9: 69385994-69385994 on Build GRCh38
rs11138396 Chr.9: 69385873-69385873 on Build GRCh38
rs754963603 Chr.9: 69385953-69385953 on Build GRCh38
rs754665330 Chr.9: 69385850-69385850 on Build GRCh38
rs1034391572 Chr.9: 69385869-69385869 on Build GRCh38
rs370008230 Chr.9: 69385878-69385878 on Build GRCh38
rs933215687 Chr.9: 69385968-69385968 on Build GRCh38
rs774785543 Chr.9: 69385901-69385901 on Build GRCh38
rs139318995 Chr.9: 69385864-69385864 on Build GRCh38
rs775956459 Chr.9: 69385909-69385909 on Build GRCh38
rs200577522 Chr.9: 69385972-69385972 on Build GRCh38
rs376622555 Chr.9: 69385977-69385977 on Build GRCh38
rs751397036 Chr.9: 69385950-69385950 on Build GRCh38
rs936834445 Chr.9: 69386017-69386017 on Build GRCh38
rs765880946 Chr.9: 69385943-69385943 on Build GRCh38
rs776663596 Chr.9: 69385971-69385971 on Build GRCh38
rs777964527 Chr.9: 69385877-69385877 on Build GRCh38
rs1026473647 Chr.9: 69385942-69385942 on Build GRCh38

View Full Product Details