Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13088689-13088962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSG1
Entrez Gene ID
83445
Gene Name
germ cell associated 1
Gene Aliases
-
UniGene
Hs.240053
Gene Chromosome Location
Chr.12: 13083537-13103720 on Build GRCh38
Transcripts
NM_001080554.2 NM_001080555.2 NM_001206842.1 NM_001206843.1 NM_001206845.1 NM_031289.3 NM_153823.3 XM_005253493.2 XM_005253495.1 XM_011520858.1 XM_011520859.1 XM_011520860.1 XM_011520861.1 XM_011520862.1 XM_011520863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1360222 c.602G>T p.G201V Chr.12: 13088741-13088741 on Build GRCh38
1360224 c.514C>T p.Q172* Chr.12: 13088829-13088829 on Build GRCh38
6418368 c.539G>T p.R180L Chr.12: 13088804-13088804 on Build GRCh38
3458208 c.328G>A p.E110K Chr.12: 13088907-13088907 on Build GRCh38
4652169 c.416G>A p.C139Y Chr.12: 13088819-13088819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143931848 Chr.12: 13088874-13088874 on Build GRCh38
rs753298883 Chr.12: 13088853-13088853 on Build GRCh38
rs938887454 Chr.12: 13088852-13088852 on Build GRCh38
rs754485360 Chr.12: 13088927-13088927 on Build GRCh38
rs766414688 Chr.12: 13088730-13088730 on Build GRCh38
rs367559691 Chr.12: 13088809-13088809 on Build GRCh38
rs762308463 Chr.12: 13088778-13088778 on Build GRCh38
rs777349642 Chr.12: 13088938-13088938 on Build GRCh38
rs759604142 Chr.12: 13088815-13088815 on Build GRCh38
rs773833859 Chr.12: 13088850-13088850 on Build GRCh38
rs755152563 Chr.12: 13088731-13088731 on Build GRCh38
rs931304646 Chr.12: 13088910-13088910 on Build GRCh38
rs747490923 Chr.12: 13088775-13088775 on Build GRCh38
rs772468902 Chr.12: 13088759-13088759 on Build GRCh38
rs144770978 Chr.12: 13088843-13088843 on Build GRCh38
rs779693983 Chr.12: 13088785-13088785 on Build GRCh38
rs770870527 Chr.12: 13088873-13088873 on Build GRCh38
rs781735511 Chr.12: 13088851-13088851 on Build GRCh38
rs965068514 Chr.12: 13088757-13088757 on Build GRCh38
rs765834004 Chr.12: 13088929-13088929 on Build GRCh38
rs753018838 Chr.12: 13088761-13088761 on Build GRCh38
rs771903046 Chr.12: 13088858-13088858 on Build GRCh38
rs763257935 Chr.12: 13088898-13088898 on Build GRCh38
rs776579379 Chr.12: 13088814-13088814 on Build GRCh38
rs764536126 Chr.12: 13088836-13088836 on Build GRCh38
rs759568046 Chr.12: 13088888-13088888 on Build GRCh38
rs764615913 Chr.12: 13088902-13088902 on Build GRCh38
rs777539932 Chr.12: 13088859-13088859 on Build GRCh38
rs554588973 Chr.12: 13088742-13088742 on Build GRCh38
rs374896777 Chr.12: 13088774-13088774 on Build GRCh38
rs773082335 Chr.12: 13088723-13088723 on Build GRCh38
rs375090125 Chr.12: 13088930-13088930 on Build GRCh38
rs758051940 Chr.12: 13088931-13088931 on Build GRCh38
rs769890340 Chr.12: 13088892-13088892 on Build GRCh38
rs760774435 Chr.12: 13088727-13088727 on Build GRCh38
rs751973707 Chr.12: 13088908-13088908 on Build GRCh38
rs768195450 Chr.12: 13088790-13088790 on Build GRCh38
rs201891084 Chr.12: 13088870-13088870 on Build GRCh38
rs776584261 Chr.12: 13088879-13088879 on Build GRCh38
rs760582393 Chr.12: 13088805-13088805 on Build GRCh38
rs749032603 Chr.12: 13088789-13088789 on Build GRCh38
rs769107461 Chr.12: 13088812-13088812 on Build GRCh38
rs778460785 Chr.12: 13088857-13088857 on Build GRCh38
rs773994830 Chr.12: 13088791-13088791 on Build GRCh38
rs954003926 Chr.12: 13088719-13088719 on Build GRCh38
rs148537880 Chr.12: 13088844-13088844 on Build GRCh38
rs747915218 Chr.12: 13088794-13088794 on Build GRCh38
rs767929695 Chr.12: 13088912-13088912 on Build GRCh38
rs1034570468 Chr.12: 13088885-13088885 on Build GRCh38
rs149406730 Chr.12: 13088833-13088833 on Build GRCh38
rs767455373 Chr.12: 13088722-13088722 on Build GRCh38
rs746741517 Chr.12: 13088865-13088865 on Build GRCh38
rs746435025 Chr.12: 13088754-13088755 on Build GRCh38
rs147266632 Chr.12: 13088897-13088897 on Build GRCh38
rs142983705 Chr.12: 13088834-13088834 on Build GRCh38
rs771812880 Chr.12: 13088800-13088800 on Build GRCh38
rs758743534 Chr.12: 13088765-13088765 on Build GRCh38
rs765401997 Chr.12: 13088825-13088825 on Build GRCh38
rs750901027 Chr.12: 13088923-13088923 on Build GRCh38
rs552857898 Chr.12: 13088909-13088909 on Build GRCh38
rs773172378 Chr.12: 13088804-13088804 on Build GRCh38

View Full Product Details