Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24445665-24445906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPLD1
Entrez Gene ID
2822
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
Gene Aliases
GPIPLD, GPIPLDM, PIGPLD, PIGPLD1, PLD
UniGene
Hs.533291
Gene Chromosome Location
Chr.6: 24425832-24495163 on Build GRCh38
Transcripts
NM_001503.3 XM_017010753.1 XM_017010754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6173536 c.1916C>G p.S639C Chr.6: 24445736-24445736 on Build GRCh38
6198157 c.1844G>A p.R615Q Chr.6: 24445808-24445808 on Build GRCh38
4473913 c.1881C>T p.Y627Y Chr.6: 24445771-24445771 on Build GRCh38
3374160 c.1849G>T p.E617* Chr.6: 24445803-24445803 on Build GRCh38
5732343 c.1843C>T p.R615* Chr.6: 24445809-24445809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549427356 Chr.6: 24445780-24445780 on Build GRCh38
rs566324510 Chr.6: 24445808-24445808 on Build GRCh38
rs761409402 Chr.6: 24445883-24445883 on Build GRCh38
rs201973866 Chr.6: 24445782-24445782 on Build GRCh38
rs367558013 Chr.6: 24445847-24445847 on Build GRCh38
rs771464586 Chr.6: 24445841-24445841 on Build GRCh38
rs760216376 Chr.6: 24445865-24445866 on Build GRCh38
rs773603377 Chr.6: 24445826-24445826 on Build GRCh38
rs370530696 Chr.6: 24445759-24445759 on Build GRCh38
rs142962906 Chr.6: 24445809-24445809 on Build GRCh38
rs769825494 Chr.6: 24445785-24445786 on Build GRCh38
rs762278395 Chr.6: 24445716-24445716 on Build GRCh38
rs780060819 Chr.6: 24445799-24445799 on Build GRCh38
rs147462718 Chr.6: 24445823-24445823 on Build GRCh38
rs762662225 Chr.6: 24445758-24445758 on Build GRCh38
rs772344726 Chr.6: 24445825-24445825 on Build GRCh38
rs779279633 Chr.6: 24445819-24445819 on Build GRCh38
rs765868702 Chr.6: 24445871-24445874 on Build GRCh38
rs761193919 Chr.6: 24445723-24445723 on Build GRCh38
rs751246675 Chr.6: 24445854-24445854 on Build GRCh38
rs757052310 Chr.6: 24445770-24445770 on Build GRCh38
rs768794956 Chr.6: 24445816-24445816 on Build GRCh38
rs536019962 Chr.6: 24445722-24445722 on Build GRCh38
rs756060393 Chr.6: 24445783-24445783 on Build GRCh38
rs375579025 Chr.6: 24445718-24445718 on Build GRCh38
rs1055670865 Chr.6: 24445856-24445856 on Build GRCh38
rs774910588 Chr.6: 24445699-24445699 on Build GRCh38
rs768991262 Chr.6: 24445697-24445697 on Build GRCh38
rs1025021090 Chr.6: 24445710-24445710 on Build GRCh38
rs761126679 Chr.6: 24445831-24445831 on Build GRCh38
rs764702231 Chr.6: 24445741-24445741 on Build GRCh38
rs752220216 Chr.6: 24445755-24445755 on Build GRCh38
rs751156763 Chr.6: 24445766-24445766 on Build GRCh38
rs112267960 Chr.6: 24445810-24445810 on Build GRCh38
rs985614161 Chr.6: 24445775-24445775 on Build GRCh38
rs952182861 Chr.6: 24445692-24445692 on Build GRCh38
rs1027664677 Chr.6: 24445757-24445757 on Build GRCh38
rs976384048 Chr.6: 24445803-24445803 on Build GRCh38
rs937145658 Chr.6: 24445855-24445855 on Build GRCh38
rs553399381 Chr.6: 24445848-24445848 on Build GRCh38

View Full Product Details