Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83837433-83837684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF27
Entrez Gene ID
55582
Gene Name
kinesin family member 27
Gene Aliases
-
UniGene
Hs.697514
Gene Chromosome Location
Chr.9: 83832795-83921539 on Build GRCh38
Transcripts
NM_001271927.1 NM_001271928.1 NM_017576.2 XM_011518848.2 XM_011518849.2 XM_011518850.2 XM_011518854.2 XM_011518857.2 XM_011518858.2 XM_017014900.1 XM_017014901.1 XM_017014902.1 XM_017014903.1 XM_017014904.1 XM_017014905.1 XM_017014906.1 XM_017014907.1 XM_017014908.1 XM_017014909.1 XM_017014910.1 XM_017014911.1 XM_017014912.1 XM_017014913.1 XM_017014915.1 XM_017014916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5632654 c.3768G>A p.M1256I Chr.9: 83837439-83837439 on Build GRCh38
4897076 c.3758G>A p.G1253E Chr.9: 83837449-83837449 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751933982 Chr.9: 83837535-83837535 on Build GRCh38
rs919485448 Chr.9: 83837468-83837468 on Build GRCh38
rs1009232940 Chr.9: 83837587-83837587 on Build GRCh38
rs965659221 Chr.9: 83837623-83837623 on Build GRCh38
rs148632153 Chr.9: 83837579-83837579 on Build GRCh38
rs953882593 Chr.9: 83837647-83837647 on Build GRCh38
rs2542780 Chr.9: 83837480-83837480 on Build GRCh38
rs866103281 Chr.9: 83837605-83837605 on Build GRCh38
rs112299613 Chr.9: 83837655-83837655 on Build GRCh38
rs779629692 Chr.9: 83837525-83837525 on Build GRCh38
rs147814329 Chr.9: 83837641-83837641 on Build GRCh38
rs905380950 Chr.9: 83837524-83837524 on Build GRCh38
rs764848149 Chr.9: 83837513-83837513 on Build GRCh38
rs1053542774 Chr.9: 83837575-83837575 on Build GRCh38
rs532802658 Chr.9: 83837519-83837519 on Build GRCh38
rs566727478 Chr.9: 83837648-83837648 on Build GRCh38
rs369903546 Chr.9: 83837515-83837515 on Build GRCh38
rs2780139 Chr.9: 83837597-83837597 on Build GRCh38
rs1055101 Chr.9: 83837539-83837539 on Build GRCh38

View Full Product Details