Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 90027563-90027828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS8
Entrez Gene ID
2622
Gene Name
growth arrest specific 8
Gene Aliases
CILD33, DRC4, GAS11
UniGene
Hs.431792
Gene Chromosome Location
Chr.16: 90019629-90044971 on Build GRCh38
Transcripts
NM_001286205.1 NM_001286208.1 NM_001286209.1 NM_001481.2 XM_005256304.4 XM_005256309.4 XM_006721175.3 XM_011522990.2 XM_011522991.1 XM_011522992.2 XM_017023122.1 XM_017023123.1 XM_017023124.1 XM_017023125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063713 c.8C>T p.P3L Chr.16: 90027640-90027640 on Build GRCh38
7100574 c.37A>C p.K13Q Chr.16: 90027669-90027669 on Build GRCh38
7283591 c.47C>T p.P16L Chr.16: 90027679-90027679 on Build GRCh38
2923977 c.86A>G p.E29G Chr.16: 90027718-90027718 on Build GRCh38
3999990 c.45C>A p.T15T Chr.16: 90027677-90027677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761578842 Chr.16: 90027620-90027620 on Build GRCh38
rs775172341 Chr.16: 90027612-90027612 on Build GRCh38
rs764817938 Chr.16: 90027693-90027693 on Build GRCh38
rs776332906 Chr.16: 90027619-90027619 on Build GRCh38
rs758352529 Chr.16: 90027650-90027650 on Build GRCh38
rs773898919 Chr.16: 90027601-90027601 on Build GRCh38
rs764469015 Chr.16: 90027773-90027773 on Build GRCh38
rs146066553 Chr.16: 90027663-90027663 on Build GRCh38
rs142699309 Chr.16: 90027708-90027708 on Build GRCh38
rs780774874 Chr.16: 90027747-90027747 on Build GRCh38
rs774208110 Chr.16: 90027760-90027760 on Build GRCh38
rs781205439 Chr.16: 90027687-90027687 on Build GRCh38
rs3743826 Chr.16: 90027806-90027806 on Build GRCh38
rs188382147 Chr.16: 90027677-90027677 on Build GRCh38
rs750407315 Chr.16: 90027640-90027640 on Build GRCh38
rs896190890 Chr.16: 90027776-90027776 on Build GRCh38
rs762671014 Chr.16: 90027642-90027642 on Build GRCh38
rs747568018 Chr.16: 90027748-90027748 on Build GRCh38
rs983274332 Chr.16: 90027622-90027622 on Build GRCh38
rs970472602 Chr.16: 90027585-90027585 on Build GRCh38
rs761502690 Chr.16: 90027692-90027692 on Build GRCh38
rs200245827 Chr.16: 90027701-90027701 on Build GRCh38
rs3743827 Chr.16: 90027787-90027787 on Build GRCh38
rs188665792 Chr.16: 90027767-90027767 on Build GRCh38
rs553992184 Chr.16: 90027791-90027791 on Build GRCh38
rs754269108 Chr.16: 90027719-90027719 on Build GRCh38
rs199840417 Chr.16: 90027734-90027734 on Build GRCh38
rs907747230 Chr.16: 90027641-90027641 on Build GRCh38
rs749792580 Chr.16: 90027608-90027608 on Build GRCh38
rs762664716 Chr.16: 90027696-90027696 on Build GRCh38
rs896377162 Chr.16: 90027606-90027606 on Build GRCh38
rs769437525 Chr.16: 90027752-90027752 on Build GRCh38
rs185349515 Chr.16: 90027686-90027686 on Build GRCh38
rs768097616 Chr.16: 90027735-90027735 on Build GRCh38
rs774941281 Chr.16: 90027680-90027680 on Build GRCh38
rs773019126 Chr.16: 90027695-90027695 on Build GRCh38
rs192874500 Chr.16: 90027679-90027679 on Build GRCh38
rs755023207 Chr.16: 90027667-90027667 on Build GRCh38
rs767318404 Chr.16: 90027766-90027766 on Build GRCh38
rs568387697 Chr.16: 90027788-90027788 on Build GRCh38
rs560531650 Chr.16: 90027717-90027717 on Build GRCh38
rs751381238 Chr.16: 90027598-90027598 on Build GRCh38
rs374981045 Chr.16: 90027699-90027699 on Build GRCh38
rs368992400 Chr.16: 90027733-90027733 on Build GRCh38
rs764984478 Chr.16: 90027632-90027632 on Build GRCh38
rs762791071 Chr.16: 90027755-90027755 on Build GRCh38
rs535373852 Chr.16: 90027789-90027789 on Build GRCh38
rs781341291 Chr.16: 90027668-90027668 on Build GRCh38
rs372771904 Chr.16: 90027597-90027597 on Build GRCh38
rs61734732 Chr.16: 90027670-90027670 on Build GRCh38
rs767716654 Chr.16: 90027729-90027730 on Build GRCh38
rs754882500 Chr.16: 90027610-90027610 on Build GRCh38
rs529831631 Chr.16: 90027752-90027753 on Build GRCh38
rs200553227 Chr.16: 90027616-90027616 on Build GRCh38
rs895400217 Chr.16: 90027785-90027785 on Build GRCh38
rs766261400 Chr.16: 90027659-90027659 on Build GRCh38
rs770684837 Chr.16: 90027757-90027757 on Build GRCh38
rs370022491 Chr.16: 90027703-90027703 on Build GRCh38
rs3743828 Chr.16: 90027761-90027761 on Build GRCh38
rs371593674 Chr.16: 90027715-90027715 on Build GRCh38
rs757684583 Chr.16: 90027727-90027727 on Build GRCh38
rs1054424948 Chr.16: 90027777-90027777 on Build GRCh38

View Full Product Details