Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30586553-30586816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G2E3
Entrez Gene ID
55632
Gene Name
G2/M-phase specific E3 ubiquitin protein ligase
Gene Aliases
KIAA1333, PHF7B
UniGene
Hs.509008
Gene Chromosome Location
Chr.14: 30559123-30620064 on Build GRCh38
Transcripts
NM_001308097.1 NM_017769.4 XM_011536925.2 XM_011536926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2034120 c.116C>T p.T39I Chr.14: 30586796-30586796 on Build GRCh38
2034117 c.52C>T p.R18* Chr.14: 30586732-30586732 on Build GRCh38
955188 c.81C>T p.Y27Y Chr.14: 30586761-30586761 on Build GRCh38
698001 c.64G>A p.D22N Chr.14: 30586744-30586744 on Build GRCh38
1369489 c.127T>C p.Y43H Chr.14: 30586807-30586807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572984978 Chr.14: 30586664-30586664 on Build GRCh38
rs755173653 Chr.14: 30586700-30586700 on Build GRCh38
rs766506513 Chr.14: 30586694-30586694 on Build GRCh38
rs753682707 Chr.14: 30586671-30586671 on Build GRCh38
rs528278657 Chr.14: 30586692-30586692 on Build GRCh38
rs1025612703 Chr.14: 30586637-30586637 on Build GRCh38
rs558033093 Chr.14: 30586748-30586748 on Build GRCh38
rs771416186 Chr.14: 30586741-30586741 on Build GRCh38
rs781238644 Chr.14: 30586713-30586713 on Build GRCh38
rs745352962 Chr.14: 30586753-30586753 on Build GRCh38
rs969598827 Chr.14: 30586603-30586603 on Build GRCh38
rs774959349 Chr.14: 30586755-30586755 on Build GRCh38
rs779858002 Chr.14: 30586768-30586770 on Build GRCh38
rs958787530 Chr.14: 30586660-30586660 on Build GRCh38
rs61734966 Chr.14: 30586778-30586778 on Build GRCh38
rs951487762 Chr.14: 30586648-30586648 on Build GRCh38
rs901394952 Chr.14: 30586611-30586611 on Build GRCh38
rs984202363 Chr.14: 30586656-30586656 on Build GRCh38
rs748449973 Chr.14: 30586725-30586725 on Build GRCh38
rs959938866 Chr.14: 30586650-30586650 on Build GRCh38
rs980239100 Chr.14: 30586614-30586614 on Build GRCh38
rs199616266 Chr.14: 30586752-30586752 on Build GRCh38
rs148066743 Chr.14: 30586761-30586761 on Build GRCh38
rs751673838 Chr.14: 30586698-30586698 on Build GRCh38
rs778230556 Chr.14: 30586739-30586739 on Build GRCh38
rs779473949 Chr.14: 30586616-30586616 on Build GRCh38
rs929859161 Chr.14: 30586663-30586664 on Build GRCh38
rs763733235 Chr.14: 30586782-30586782 on Build GRCh38
rs774906986 Chr.14: 30586750-30586750 on Build GRCh38
rs758209735 Chr.14: 30586697-30586700 on Build GRCh38
rs563987859 Chr.14: 30586613-30586613 on Build GRCh38
rs370229233 Chr.14: 30586733-30586733 on Build GRCh38
rs1017470779 Chr.14: 30586658-30586658 on Build GRCh38

View Full Product Details