Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15888429-15888686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOPNL
Entrez Gene ID
123811
Gene Name
FGFR1OP N-terminal like
Gene Aliases
C16orf63, FOR20, PHSECRG2
UniGene
Hs.514179
Gene Chromosome Location
Chr.16: 15865719-15888649 on Build GRCh38
Transcripts
NM_001304497.1 NM_001304498.1 NM_001304499.1 NM_001304500.1 NM_001304502.1 NM_144600.3 NR_130755.1 NR_130756.1 NR_130757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619058 c.16G>T p.E6* Chr.16: 15888570-15888570 on Build GRCh38
5635103 c.24G>A p.K8K Chr.16: 15888562-15888562 on Build GRCh38
6666009 c.5C>T p.A2V Chr.16: 15888581-15888581 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756830363 Chr.16: 15888557-15888557 on Build GRCh38
rs950957628 Chr.16: 15888620-15888620 on Build GRCh38
rs772924623 Chr.16: 15888604-15888611 on Build GRCh38
rs767726226 Chr.16: 15888608-15888610 on Build GRCh38
rs755666077 Chr.16: 15888541-15888541 on Build GRCh38
rs781261407 Chr.16: 15888603-15888603 on Build GRCh38
rs145163100 Chr.16: 15888491-15888491 on Build GRCh38
rs753574309 Chr.16: 15888589-15888589 on Build GRCh38
rs572715049 Chr.16: 15888538-15888538 on Build GRCh38
rs533601912 Chr.16: 15888594-15888594 on Build GRCh38
rs145630355 Chr.16: 15888579-15888579 on Build GRCh38
rs563175476 Chr.16: 15888585-15888585 on Build GRCh38
rs377757838 Chr.16: 15888545-15888545 on Build GRCh38
rs370143085 Chr.16: 15888597-15888597 on Build GRCh38
rs758915210 Chr.16: 15888595-15888595 on Build GRCh38
rs1009673624 Chr.16: 15888535-15888535 on Build GRCh38
rs373908219 Chr.16: 15888604-15888604 on Build GRCh38
rs755421719 Chr.16: 15888602-15888602 on Build GRCh38
rs767367770 Chr.16: 15888539-15888539 on Build GRCh38
rs1017465300 Chr.16: 15888572-15888572 on Build GRCh38
rs557000485 Chr.16: 15888654-15888654 on Build GRCh38
rs112250647 Chr.16: 15888622-15888622 on Build GRCh38
rs138233634 Chr.16: 15888616-15888616 on Build GRCh38
rs775219497 Chr.16: 15888581-15888581 on Build GRCh38
rs746987047 Chr.16: 15888465-15888465 on Build GRCh38
rs776142967 Chr.16: 15888626-15888626 on Build GRCh38
rs768924843 Chr.16: 15888530-15888530 on Build GRCh38
rs756708724 Chr.16: 15888591-15888591 on Build GRCh38
rs114523129 Chr.16: 15888632-15888632 on Build GRCh38
rs1044602963 Chr.16: 15888613-15888613 on Build GRCh38
rs373761532 Chr.16: 15888571-15888571 on Build GRCh38
rs371449713 Chr.16: 15888560-15888560 on Build GRCh38
rs373618119 Chr.16: 15888628-15888628 on Build GRCh38
rs377297363 Chr.16: 15888569-15888569 on Build GRCh38
rs772557013 Chr.16: 15888562-15888562 on Build GRCh38
rs140526629 Chr.16: 15888531-15888531 on Build GRCh38
rs752890102 Chr.16: 15888610-15888638 on Build GRCh38
rs200386485 Chr.16: 15888567-15888567 on Build GRCh38
rs114079614 Chr.16: 15888537-15888537 on Build GRCh38
rs965838634 Chr.16: 15888573-15888573 on Build GRCh38
rs370555988 Chr.16: 15888574-15888574 on Build GRCh38
rs377654029 Chr.16: 15888523-15888523 on Build GRCh38
rs370768074 Chr.16: 15888635-15888635 on Build GRCh38
rs770009442 Chr.16: 15888610-15888610 on Build GRCh38
rs201357085 Chr.16: 15888611-15888611 on Build GRCh38
rs746434262 Chr.16: 15888561-15888561 on Build GRCh38
rs760868898 Chr.16: 15888524-15888524 on Build GRCh38
rs909493060 Chr.16: 15888475-15888475 on Build GRCh38
rs769789210 Chr.16: 15888543-15888560 on Build GRCh38
rs574682170 Chr.16: 15888471-15888471 on Build GRCh38
rs899209317 Chr.16: 15888615-15888615 on Build GRCh38
rs112502876 Chr.16: 15888516-15888516 on Build GRCh38
rs762750257 Chr.16: 15888604-15888627 on Build GRCh38
rs773044024 Chr.16: 15888649-15888649 on Build GRCh38
rs1018211410 Chr.16: 15888515-15888515 on Build GRCh38
rs771958235 Chr.16: 15888612-15888612 on Build GRCh38
rs760551805 Chr.16: 15888584-15888584 on Build GRCh38
rs200435974 Chr.16: 15888617-15888617 on Build GRCh38
rs557211036 Chr.16: 15888499-15888499 on Build GRCh38
rs1026575606 Chr.16: 15888526-15888526 on Build GRCh38
rs752494468 Chr.16: 15888540-15888540 on Build GRCh38
rs776955193 Chr.16: 15888570-15888570 on Build GRCh38
rs113574900 Chr.16: 15888533-15888533 on Build GRCh38
rs529279150 Chr.16: 15888578-15888578 on Build GRCh38
rs774271736 Chr.16: 15888576-15888576 on Build GRCh38
rs747308287 Chr.16: 15888599-15888599 on Build GRCh38
rs903494016 Chr.16: 15888648-15888648 on Build GRCh38
rs760993749 Chr.16: 15888613-15888619 on Build GRCh38
rs764906306 Chr.16: 15888593-15888593 on Build GRCh38
rs746201336 Chr.16: 15888513-15888513 on Build GRCh38
rs1049039579 Chr.16: 15888450-15888450 on Build GRCh38
rs373355444 Chr.16: 15888655-15888655 on Build GRCh38
rs780495622 Chr.16: 15888565-15888565 on Build GRCh38
rs766198527 Chr.16: 15888536-15888536 on Build GRCh38
rs1009312381 Chr.16: 15888629-15888629 on Build GRCh38
rs766043843 Chr.16: 15888556-15888556 on Build GRCh38

View Full Product Details