Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45340273-45340514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM118A
Entrez Gene ID
55007
Gene Name
family with sequence similarity 118 member A
Gene Aliases
C22orf8
UniGene
Hs.265018
Gene Chromosome Location
Chr.22: 45309200-45341955 on Build GRCh38
Transcripts
NM_001104595.1 NM_017911.2 XM_011530247.2 XM_011530254.2 XM_017028841.1 XM_017028842.1 XM_017028843.1 XM_017028844.1 XM_017028845.1 XM_017028846.1 XM_017028847.1 XM_017028848.1 XM_017028849.1 XM_017028850.1 XM_017028851.1 XM_017028852.1 XM_017028854.1 XM_017028861.1

SNP Detail

SNP ID Chromosome Location
rs778317540 Chr.22: 45340442-45340442 on Build GRCh38
rs144465524 Chr.22: 45340446-45340446 on Build GRCh38
rs771564295 Chr.22: 45340303-45340303 on Build GRCh38
rs574613187 Chr.22: 45340329-45340329 on Build GRCh38
rs765325484 Chr.22: 45340395-45340395 on Build GRCh38
rs746072385 Chr.22: 45340448-45340448 on Build GRCh38
rs760338537 Chr.22: 45340356-45340356 on Build GRCh38
rs774396192 Chr.22: 45340447-45340447 on Build GRCh38
rs899062567 Chr.22: 45340304-45340304 on Build GRCh38
rs770269926 Chr.22: 45340440-45340440 on Build GRCh38
rs753727926 Chr.22: 45340372-45340372 on Build GRCh38
rs771053175 Chr.22: 45340352-45340352 on Build GRCh38
rs952983729 Chr.22: 45340325-45340325 on Build GRCh38
rs749951892 Chr.22: 45340398-45340398 on Build GRCh38
rs749258607 Chr.22: 45340444-45340444 on Build GRCh38
rs764013044 Chr.22: 45340366-45340366 on Build GRCh38
rs534944811 Chr.22: 45340465-45340465 on Build GRCh38
rs748558827 Chr.22: 45340428-45340428 on Build GRCh38
rs372325274 Chr.22: 45340313-45340313 on Build GRCh38
rs765560219 Chr.22: 45340446-45340446 on Build GRCh38
rs774637707 Chr.22: 45340354-45340354 on Build GRCh38
rs934606246 Chr.22: 45340491-45340491 on Build GRCh38
rs149231997 Chr.22: 45340376-45340376 on Build GRCh38
rs139521541 Chr.22: 45340320-45340320 on Build GRCh38
rs554594993 Chr.22: 45340326-45340326 on Build GRCh38
rs781502525 Chr.22: 45340422-45340422 on Build GRCh38
rs371436250 Chr.22: 45340414-45340414 on Build GRCh38
rs915696369 Chr.22: 45340333-45340333 on Build GRCh38
rs978647783 Chr.22: 45340459-45340459 on Build GRCh38
rs375088388 Chr.22: 45340466-45340466 on Build GRCh38
rs751243050 Chr.22: 45340409-45340409 on Build GRCh38
rs779518939 Chr.22: 45340406-45340406 on Build GRCh38
rs12167007 Chr.22: 45340337-45340337 on Build GRCh38
rs772473159 Chr.22: 45340450-45340450 on Build GRCh38
rs1053473921 Chr.22: 45340492-45340492 on Build GRCh38

View Full Product Details