Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1474690-1474961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS1
Entrez Gene ID
26173
Gene Name
integrator complex subunit 1
Gene Aliases
INT1, NET28
UniGene
Hs.532188
Gene Chromosome Location
Chr.7: 1470277-1504382 on Build GRCh38
Transcripts
NM_001080453.2 XM_011515260.1 XM_017011959.1 XM_017011960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3698280 c.6225G>A p.V2075V Chr.7: 1474728-1474728 on Build GRCh38
4161988 c.6129C>A p.I2043I Chr.7: 1474824-1474824 on Build GRCh38
5749061 c.6223G>T p.V2075L Chr.7: 1474730-1474730 on Build GRCh38
6686377 c.6133C>T p.R2045C Chr.7: 1474820-1474820 on Build GRCh38
6504574 c.6154G>A p.D2052N Chr.7: 1474799-1474799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931147837 Chr.7: 1474920-1474920 on Build GRCh38
rs1039185602 Chr.7: 1474846-1474846 on Build GRCh38
rs753239248 Chr.7: 1474725-1474725 on Build GRCh38
rs375559240 Chr.7: 1474853-1474853 on Build GRCh38
rs374803757 Chr.7: 1474742-1474742 on Build GRCh38
rs1040752382 Chr.7: 1474933-1474933 on Build GRCh38
rs780284734 Chr.7: 1474819-1474819 on Build GRCh38
rs370450469 Chr.7: 1474768-1474768 on Build GRCh38
rs1026099894 Chr.7: 1474910-1474910 on Build GRCh38
rs755064171 Chr.7: 1474832-1474832 on Build GRCh38
rs868140630 Chr.7: 1474756-1474756 on Build GRCh38
rs756207848 Chr.7: 1474762-1474762 on Build GRCh38
rs901937575 Chr.7: 1474935-1474935 on Build GRCh38
rs973071489 Chr.7: 1474814-1474814 on Build GRCh38
rs946808454 Chr.7: 1474852-1474852 on Build GRCh38
rs752120836 Chr.7: 1474808-1474808 on Build GRCh38
rs374496492 Chr.7: 1474810-1474810 on Build GRCh38
rs759166691 Chr.7: 1474790-1474790 on Build GRCh38
rs950442951 Chr.7: 1474822-1474822 on Build GRCh38
rs182321860 Chr.7: 1474875-1474875 on Build GRCh38
rs994983848 Chr.7: 1474901-1474901 on Build GRCh38
rs772578709 Chr.7: 1474844-1474844 on Build GRCh38
rs757579147 Chr.7: 1474736-1474736 on Build GRCh38
rs1039091955 Chr.7: 1474717-1474717 on Build GRCh38
rs746759664 Chr.7: 1474783-1474783 on Build GRCh38
rs559688069 Chr.7: 1474731-1474731 on Build GRCh38
rs561160944 Chr.7: 1474809-1474809 on Build GRCh38
rs368503457 Chr.7: 1474820-1474820 on Build GRCh38
rs140491630 Chr.7: 1474869-1474869 on Build GRCh38
rs11542325 Chr.7: 1474734-1474734 on Build GRCh38
rs373812497 Chr.7: 1474780-1474780 on Build GRCh38
rs919717425 Chr.7: 1474918-1474918 on Build GRCh38
rs146843925 Chr.7: 1474864-1474864 on Build GRCh38
rs1057066447 Chr.7: 1474865-1474865 on Build GRCh38
rs368079404 Chr.7: 1474856-1474856 on Build GRCh38
rs551948401 Chr.7: 1474941-1474941 on Build GRCh38
rs376092004 Chr.7: 1474761-1474761 on Build GRCh38
rs950431858 Chr.7: 1474932-1474932 on Build GRCh38
rs190550247 Chr.7: 1474724-1474724 on Build GRCh38
rs553447758 Chr.7: 1474784-1474784 on Build GRCh38
rs373798063 Chr.7: 1474773-1474773 on Build GRCh38
rs915299492 Chr.7: 1474849-1474849 on Build GRCh38
rs1022824404 Chr.7: 1474878-1474878 on Build GRCh38
rs201606213 Chr.7: 1474801-1474801 on Build GRCh38
rs985380035 Chr.7: 1474922-1474922 on Build GRCh38
rs572103436 Chr.7: 1474800-1474800 on Build GRCh38
rs116121226 Chr.7: 1474728-1474728 on Build GRCh38
rs763765156 Chr.7: 1474877-1474877 on Build GRCh38
rs751928685 Chr.7: 1474732-1474732 on Build GRCh38
rs963267911 Chr.7: 1474880-1474880 on Build GRCh38
rs771797708 Chr.7: 1474817-1474817 on Build GRCh38
rs987675580 Chr.7: 1474825-1474825 on Build GRCh38
rs748045769 Chr.7: 1474779-1474779 on Build GRCh38
rs961390757 Chr.7: 1474907-1474907 on Build GRCh38
rs911122682 Chr.7: 1474923-1474923 on Build GRCh38
rs764642443 Chr.7: 1474805-1474805 on Build GRCh38
rs1012759461 Chr.7: 1474795-1474795 on Build GRCh38
rs372112448 Chr.7: 1474833-1474833 on Build GRCh38
rs371995859 Chr.7: 1474863-1474863 on Build GRCh38
rs766050980 Chr.7: 1474918-1474918 on Build GRCh38
rs776338733 Chr.7: 1474785-1474785 on Build GRCh38
rs367655126 Chr.7: 1474791-1474791 on Build GRCh38

View Full Product Details