Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112423819-112424078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK1
Entrez Gene ID
80216
Gene Name
alpha kinase 1
Gene Aliases
8430410J10Rik, LAK
UniGene
Hs.652825
Gene Chromosome Location
Chr.4: 112286420-112442618 on Build GRCh38
Transcripts
NM_001102406.1 NM_001253884.1 NM_025144.3 XM_005263245.4 XM_005263246.4 XM_006714326.3 XM_017008633.1 XM_017008634.1 XM_017008635.1 XM_017008636.1 XM_017008637.1 XM_017008638.1 XM_017008639.1 XM_017008640.1 XM_017008641.1 XM_017008642.1 XM_017008643.1 XM_017008644.1 XM_017008645.1 XM_017008646.1 XM_017008647.1 XM_017008648.1 XM_017008649.1 XM_017008650.1 XM_017008651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1050104 c.484T>G p.L162V Chr.4: 112423952-112423952 on Build GRCh38
1050106 c.505A>G p.S169G Chr.4: 112423973-112423973 on Build GRCh38
4657329 c.521A>G p.N174S Chr.4: 112423989-112423989 on Build GRCh38
5466020 c.517A>C p.S173R Chr.4: 112423985-112423985 on Build GRCh38
3736330 c.523A>G p.N175D Chr.4: 112423991-112423991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984503890 Chr.4: 112423926-112423926 on Build GRCh38
rs753884499 Chr.4: 112424046-112424046 on Build GRCh38
rs1013335065 Chr.4: 112423877-112423877 on Build GRCh38
rs538240091 Chr.4: 112423885-112423885 on Build GRCh38
rs372294966 Chr.4: 112423899-112423899 on Build GRCh38
rs201681617 Chr.4: 112423936-112423936 on Build GRCh38
rs138427105 Chr.4: 112423957-112423957 on Build GRCh38
rs556622026 Chr.4: 112423886-112423886 on Build GRCh38
rs772824878 Chr.4: 112423982-112423982 on Build GRCh38
rs764210870 Chr.4: 112424029-112424029 on Build GRCh38
rs768931876 Chr.4: 112423910-112423910 on Build GRCh38
rs889658899 Chr.4: 112423948-112423948 on Build GRCh38
rs760770272 Chr.4: 112424027-112424027 on Build GRCh38
rs746240245 Chr.4: 112423924-112423924 on Build GRCh38
rs6533616 Chr.4: 112423991-112423991 on Build GRCh38
rs764333492 Chr.4: 112423906-112423906 on Build GRCh38
rs757452670 Chr.4: 112423919-112423919 on Build GRCh38
rs763571551 Chr.4: 112423902-112423902 on Build GRCh38
rs762182103 Chr.4: 112423945-112423945 on Build GRCh38
rs767473198 Chr.4: 112423898-112423898 on Build GRCh38
rs774069152 Chr.4: 112424001-112424001 on Build GRCh38
rs769379637 Chr.4: 112423970-112423970 on Build GRCh38
rs759453594 Chr.4: 112423896-112423896 on Build GRCh38
rs923826311 Chr.4: 112423876-112423876 on Build GRCh38
rs773792570 Chr.4: 112423890-112423890 on Build GRCh38
rs542475979 Chr.4: 112423859-112423859 on Build GRCh38
rs376769752 Chr.4: 112424002-112424002 on Build GRCh38
rs775209151 Chr.4: 112424007-112424007 on Build GRCh38
rs111446125 Chr.4: 112423904-112423904 on Build GRCh38
rs770698564 Chr.4: 112423995-112423995 on Build GRCh38
rs779290867 Chr.4: 112423922-112423922 on Build GRCh38
rs754014383 Chr.4: 112423911-112423911 on Build GRCh38
rs374746088 Chr.4: 112423892-112423892 on Build GRCh38
rs776989166 Chr.4: 112423936-112423940 on Build GRCh38
rs767363240 Chr.4: 112424016-112424016 on Build GRCh38

View Full Product Details