Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159971913-159972158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA1B
Entrez Gene ID
147
Gene Name
adrenoceptor alpha 1B
Gene Aliases
ADRA1, ALPHA1BAR
UniGene
Hs.368632
Gene Chromosome Location
Chr.5: 159916733-159989205 on Build GRCh38
Transcripts
NM_000679.3 XM_006714821.3 XM_011534435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
736968 c.1054A>C p.S352R Chr.5: 159971983-159971983 on Build GRCh38
1065149 c.1026C>A p.C342* Chr.5: 159971955-159971955 on Build GRCh38
265971 c.1070G>A p.R357H Chr.5: 159971999-159971999 on Build GRCh38
7278023 c.1168C>T p.P390S Chr.5: 159972097-159972097 on Build GRCh38
3697110 c.1095C>T p.C365C Chr.5: 159972024-159972024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777674763 Chr.5: 159971980-159971980 on Build GRCh38
rs753401537 Chr.5: 159972029-159972055 on Build GRCh38
rs778237365 Chr.5: 159972129-159972129 on Build GRCh38
rs758021350 Chr.5: 159972040-159972041 on Build GRCh38
rs369546656 Chr.5: 159972004-159972004 on Build GRCh38
rs754379100 Chr.5: 159972031-159972051 on Build GRCh38
rs989832048 Chr.5: 159972107-159972107 on Build GRCh38
rs8192450 Chr.5: 159972045-159972045 on Build GRCh38
rs759092567 Chr.5: 159972031-159972031 on Build GRCh38
rs780937670 Chr.5: 159972045-159972046 on Build GRCh38
rs749687924 Chr.5: 159972136-159972136 on Build GRCh38
rs931846104 Chr.5: 159972134-159972134 on Build GRCh38
rs758441189 Chr.5: 159971983-159971983 on Build GRCh38
rs770175307 Chr.5: 159972125-159972125 on Build GRCh38
rs775699892 Chr.5: 159972052-159972063 on Build GRCh38
rs767862161 Chr.5: 159972029-159972052 on Build GRCh38
rs532690456 Chr.5: 159972060-159972060 on Build GRCh38
rs752400706 Chr.5: 159972035-159972043 on Build GRCh38
rs933314987 Chr.5: 159972007-159972007 on Build GRCh38
rs745969860 Chr.5: 159972051-159972052 on Build GRCh38
rs767215435 Chr.5: 159971962-159971962 on Build GRCh38
rs763518191 Chr.5: 159972025-159972025 on Build GRCh38
rs763916062 Chr.5: 159972050-159972050 on Build GRCh38
rs763264015 Chr.5: 159971987-159971989 on Build GRCh38
rs755149691 Chr.5: 159972066-159972066 on Build GRCh38
rs780067610 Chr.5: 159971985-159971985 on Build GRCh38
rs764612121 Chr.5: 159972023-159972043 on Build GRCh38
rs770336712 Chr.5: 159972011-159972011 on Build GRCh38
rs959165871 Chr.5: 159971999-159971999 on Build GRCh38
rs757243904 Chr.5: 159972043-159972051 on Build GRCh38
rs376920965 Chr.5: 159971968-159971968 on Build GRCh38
rs773829274 Chr.5: 159972019-159972019 on Build GRCh38
rs760441080 Chr.5: 159972046-159972046 on Build GRCh38
rs781253324 Chr.5: 159972088-159972088 on Build GRCh38
rs771591500 Chr.5: 159972027-159972027 on Build GRCh38
rs201375864 Chr.5: 159972030-159972030 on Build GRCh38
rs957670819 Chr.5: 159972097-159972097 on Build GRCh38
rs781479331 Chr.5: 159972000-159972000 on Build GRCh38
rs757136411 Chr.5: 159972053-159972053 on Build GRCh38
rs764718960 Chr.5: 159972035-159972052 on Build GRCh38
rs748447792 Chr.5: 159972105-159972105 on Build GRCh38
rs921863808 Chr.5: 159972114-159972114 on Build GRCh38
rs762411639 Chr.5: 159972028-159972029 on Build GRCh38
rs754540841 Chr.5: 159971959-159971959 on Build GRCh38
rs778832771 Chr.5: 159972055-159972055 on Build GRCh38
rs764801657 Chr.5: 159972042-159972043 on Build GRCh38
rs767073094 Chr.5: 159972036-159972036 on Build GRCh38
rs911978057 Chr.5: 159972140-159972140 on Build GRCh38
rs774836854 Chr.5: 159972029-159972034 on Build GRCh38
rs552601158 Chr.5: 159972102-159972102 on Build GRCh38
rs749273189 Chr.5: 159971981-159971981 on Build GRCh38
rs770066678 Chr.5: 159972052-159972054 on Build GRCh38
rs200619403 Chr.5: 159971998-159971998 on Build GRCh38
rs752370719 Chr.5: 159971966-159971966 on Build GRCh38
rs777582523 Chr.5: 159972041-159972058 on Build GRCh38

View Full Product Details