Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38318194-38318455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG10B
Entrez Gene ID
144245
Gene Name
ALG10B, alpha-1,2-glucosyltransferase
Gene Aliases
ALG10, KCR1
UniGene
Hs.259305
Gene Chromosome Location
Chr.12: 38316679-38329726 on Build GRCh38
Transcripts
NM_001013620.3 NM_001308340.1 XM_005268665.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4959916 c.200G>A p.G67D Chr.12: 38318289-38318289 on Build GRCh38
938889 c.286C>T p.L96F Chr.12: 38318375-38318375 on Build GRCh38
4803339 c.235G>A p.A79T Chr.12: 38318324-38318324 on Build GRCh38
431046 c.283A>C p.M95L Chr.12: 38318372-38318372 on Build GRCh38
2206499 c.332T>A p.L111Q Chr.12: 38318421-38318421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771251907 Chr.12: 38318336-38318336 on Build GRCh38
rs773865389 Chr.12: 38318395-38318395 on Build GRCh38
rs774059168 Chr.12: 38318274-38318274 on Build GRCh38
rs368182821 Chr.12: 38318257-38318257 on Build GRCh38
rs777814298 Chr.12: 38318359-38318359 on Build GRCh38
rs62001043 Chr.12: 38318315-38318315 on Build GRCh38
rs749459615 Chr.12: 38318238-38318238 on Build GRCh38
rs374009031 Chr.12: 38318351-38318351 on Build GRCh38
rs755843656 Chr.12: 38318367-38318367 on Build GRCh38
rs757173725 Chr.12: 38318239-38318239 on Build GRCh38
rs141713734 Chr.12: 38318281-38318281 on Build GRCh38
rs139511694 Chr.12: 38318352-38318352 on Build GRCh38
rs898487675 Chr.12: 38318296-38318296 on Build GRCh38
rs780082142 Chr.12: 38318377-38318377 on Build GRCh38
rs61730289 Chr.12: 38318418-38318418 on Build GRCh38
rs754266717 Chr.12: 38318307-38318307 on Build GRCh38
rs966887909 Chr.12: 38318403-38318403 on Build GRCh38
rs781050613 Chr.12: 38318241-38318241 on Build GRCh38
rs6582584 Chr.12: 38318340-38318340 on Build GRCh38
rs979204635 Chr.12: 38318405-38318405 on Build GRCh38
rs767099870 Chr.12: 38318289-38318289 on Build GRCh38
rs768214614 Chr.12: 38318387-38318387 on Build GRCh38
rs765768323 Chr.12: 38318421-38318421 on Build GRCh38
rs760899298 Chr.12: 38318299-38318299 on Build GRCh38
rs763081108 Chr.12: 38318429-38318429 on Build GRCh38
rs769804736 Chr.12: 38318243-38318243 on Build GRCh38
rs771733315 Chr.12: 38318401-38318401 on Build GRCh38
rs147838361 Chr.12: 38318248-38318257 on Build GRCh38
rs748881222 Chr.12: 38318251-38318251 on Build GRCh38
rs760468440 Chr.12: 38318400-38318400 on Build GRCh38
rs755173877 Chr.12: 38318319-38318319 on Build GRCh38
rs62001042 Chr.12: 38318306-38318306 on Build GRCh38
rs772717701 Chr.12: 38318402-38318402 on Build GRCh38
rs1047572310 Chr.12: 38318233-38318233 on Build GRCh38
rs773632002 Chr.12: 38318293-38318293 on Build GRCh38
rs749220164 Chr.12: 38318381-38318381 on Build GRCh38
rs761140434 Chr.12: 38318409-38318409 on Build GRCh38
rs764618741 Chr.12: 38318318-38318319 on Build GRCh38
rs142306686 Chr.12: 38318424-38318424 on Build GRCh38
rs745701002 Chr.12: 38318364-38318364 on Build GRCh38
rs879249621 Chr.12: 38318273-38318273 on Build GRCh38
rs771194076 Chr.12: 38318412-38318412 on Build GRCh38
rs777249616 Chr.12: 38318413-38318413 on Build GRCh38
rs745579610 Chr.12: 38318242-38318242 on Build GRCh38
rs144238652 Chr.12: 38318399-38318399 on Build GRCh38

View Full Product Details