Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132559752-132560013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD11
Entrez Gene ID
84129
Gene Name
acyl-CoA dehydrogenase family member 11
Gene Aliases
ACAD-11
UniGene
Hs.441378
Gene Chromosome Location
Chr.3: 132558138-132660131 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NR_037804.1

Gene Symbol
NPHP3-ACAD11
Entrez Gene ID
100532724
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.511991
Gene Chromosome Location
Chr.3: 132558138-132722459 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NR_037804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369227 c.2164G>A p.V722I Chr.3: 132559897-132559897 on Build GRCh38
4775711 c.2216C>T p.P739L Chr.3: 132559845-132559845 on Build GRCh38
1616933 c.2151T>C p.A717A Chr.3: 132559910-132559910 on Build GRCh38
1730580 c.2141C>G p.A714G Chr.3: 132559920-132559920 on Build GRCh38
158839 c.2146C>T p.R716W Chr.3: 132559915-132559915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037010463 Chr.3: 132559807-132559807 on Build GRCh38
rs764063988 Chr.3: 132559845-132559846 on Build GRCh38
rs755759202 Chr.3: 132559916-132559916 on Build GRCh38
rs780713021 Chr.3: 132559846-132559846 on Build GRCh38
rs921474955 Chr.3: 132559980-132559980 on Build GRCh38
rs980655233 Chr.3: 132559800-132559800 on Build GRCh38
rs932827264 Chr.3: 132559981-132559981 on Build GRCh38
rs527824466 Chr.3: 132559969-132559969 on Build GRCh38
rs762360085 Chr.3: 132559783-132559783 on Build GRCh38
rs966047891 Chr.3: 132559965-132559965 on Build GRCh38
rs749232806 Chr.3: 132559915-132559915 on Build GRCh38
rs200528059 Chr.3: 132559874-132559874 on Build GRCh38
rs763026531 Chr.3: 132559972-132559972 on Build GRCh38
rs772683533 Chr.3: 132559883-132559883 on Build GRCh38
rs779759456 Chr.3: 132559917-132559917 on Build GRCh38
rs753707771 Chr.3: 132559855-132559855 on Build GRCh38
rs777026457 Chr.3: 132559949-132559949 on Build GRCh38
rs562348380 Chr.3: 132559799-132559799 on Build GRCh38
rs943563406 Chr.3: 132559844-132559845 on Build GRCh38
rs748702983 Chr.3: 132559878-132559878 on Build GRCh38
rs1035564215 Chr.3: 132559887-132559887 on Build GRCh38
rs765277247 Chr.3: 132559901-132559901 on Build GRCh38
rs992269490 Chr.3: 132559788-132559788 on Build GRCh38
rs770046978 Chr.3: 132559958-132559958 on Build GRCh38
rs777596834 Chr.3: 132559811-132559811 on Build GRCh38
rs766694470 Chr.3: 132559889-132559889 on Build GRCh38
rs1040799814 Chr.3: 132559890-132559890 on Build GRCh38
rs776728587 Chr.3: 132559896-132559896 on Build GRCh38
rs1012721691 Chr.3: 132559908-132559908 on Build GRCh38
rs745467475 Chr.3: 132559872-132559872 on Build GRCh38
rs771218815 Chr.3: 132559945-132559945 on Build GRCh38
rs751522419 Chr.3: 132559988-132559988 on Build GRCh38
rs746794850 Chr.3: 132559820-132559820 on Build GRCh38
rs752756095 Chr.3: 132559904-132559904 on Build GRCh38
rs199545368 Chr.3: 132559920-132559920 on Build GRCh38
rs756972848 Chr.3: 132559827-132559827 on Build GRCh38
rs757017604 Chr.3: 132559914-132559914 on Build GRCh38
rs377291697 Chr.3: 132559792-132559792 on Build GRCh38
rs879069659 Chr.3: 132559852-132559852 on Build GRCh38
rs778815788 Chr.3: 132559967-132559968 on Build GRCh38
rs529367267 Chr.3: 132559898-132559898 on Build GRCh38
rs757413344 Chr.3: 132559802-132559802 on Build GRCh38
rs370802390 Chr.3: 132559838-132559838 on Build GRCh38
rs1031321398 Chr.3: 132559940-132559940 on Build GRCh38
rs757077637 Chr.3: 132559909-132559910 on Build GRCh38
rs764105784 Chr.3: 132559976-132559976 on Build GRCh38
rs376472101 Chr.3: 132559796-132559796 on Build GRCh38
rs968474175 Chr.3: 132559931-132559931 on Build GRCh38
rs761113325 Chr.3: 132559885-132559885 on Build GRCh38
rs772769438 Chr.3: 132559921-132559921 on Build GRCh38
rs766493596 Chr.3: 132559793-132559793 on Build GRCh38
rs768024945 Chr.3: 132559791-132559791 on Build GRCh38
rs931094546 Chr.3: 132559824-132559824 on Build GRCh38
rs747458128 Chr.3: 132559936-132559936 on Build GRCh38
rs752666302 Chr.3: 132559803-132559803 on Build GRCh38
rs760532647 Chr.3: 132559795-132559795 on Build GRCh38
rs377557043 Chr.3: 132559974-132559974 on Build GRCh38
rs374672178 Chr.3: 132559884-132559884 on Build GRCh38
rs758391983 Chr.3: 132559909-132559909 on Build GRCh38
rs764015856 Chr.3: 132559913-132559913 on Build GRCh38
rs886556111 Chr.3: 132559874-132559875 on Build GRCh38
rs769209249 Chr.3: 132559873-132559873 on Build GRCh38

View Full Product Details