Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68482704-68482941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC1
Entrez Gene ID
55635
Gene Name
DEP domain containing 1
Gene Aliases
DEP.8, DEPDC1-V2, DEPDC1A, SDP35
UniGene
Hs.445098
Gene Chromosome Location
Chr.1: 68474152-68497221 on Build GRCh38
Transcripts
NM_001114120.2 NM_017779.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3400985 c.1077A>G p.S359S Chr.1: 68482731-68482731 on Build GRCh38
6289805 c.1026T>G p.T342T Chr.1: 68482782-68482782 on Build GRCh38
4186064 c.1080T>G p.D360E Chr.1: 68482728-68482728 on Build GRCh38
4677066 c.1055G>C p.R352T Chr.1: 68482753-68482753 on Build GRCh38
3805859 c.1017C>G p.F339L Chr.1: 68482791-68482791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781462753 Chr.1: 68482775-68482775 on Build GRCh38
rs1014163038 Chr.1: 68482818-68482818 on Build GRCh38
rs776299020 Chr.1: 68482768-68482770 on Build GRCh38
rs898719565 Chr.1: 68482823-68482823 on Build GRCh38
rs202172428 Chr.1: 68482857-68482857 on Build GRCh38
rs775075655 Chr.1: 68482879-68482879 on Build GRCh38
rs768423262 Chr.1: 68482743-68482744 on Build GRCh38
rs996170214 Chr.1: 68482828-68482828 on Build GRCh38
rs760464782 Chr.1: 68482744-68482744 on Build GRCh38
rs763234475 Chr.1: 68482768-68482768 on Build GRCh38
rs776058498 Chr.1: 68482734-68482736 on Build GRCh38
rs757817887 Chr.1: 68482772-68482772 on Build GRCh38
rs758984738 Chr.1: 68482865-68482865 on Build GRCh38
rs999866879 Chr.1: 68482751-68482751 on Build GRCh38
rs764822761 Chr.1: 68482736-68482736 on Build GRCh38
rs777517396 Chr.1: 68482844-68482844 on Build GRCh38
rs762229351 Chr.1: 68482893-68482893 on Build GRCh38
rs764516175 Chr.1: 68482769-68482769 on Build GRCh38
rs749520646 Chr.1: 68482800-68482800 on Build GRCh38
rs752340706 Chr.1: 68482762-68482762 on Build GRCh38
rs771817464 Chr.1: 68482822-68482822 on Build GRCh38
rs780143784 Chr.1: 68482792-68482792 on Build GRCh38
rs752170095 Chr.1: 68482771-68482771 on Build GRCh38
rs775120941 Chr.1: 68482738-68482739 on Build GRCh38
rs530887977 Chr.1: 68482901-68482901 on Build GRCh38
rs750552604 Chr.1: 68482784-68482784 on Build GRCh38
rs780526610 Chr.1: 68482900-68482900 on Build GRCh38
rs769286335 Chr.1: 68482872-68482872 on Build GRCh38
rs563988599 Chr.1: 68482852-68482852 on Build GRCh38
rs761606836 Chr.1: 68482854-68482860 on Build GRCh38
rs201449469 Chr.1: 68482884-68482884 on Build GRCh38
rs756159027 Chr.1: 68482789-68482789 on Build GRCh38
rs762654774 Chr.1: 68482882-68482882 on Build GRCh38

View Full Product Details