Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123137168-123137423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMX3
Entrez Gene ID
340784
Gene Name
H6 family homeobox 3
Gene Aliases
NKX-5.1, NKX5.1, Nkx5-1
UniGene
Hs.531194
Gene Chromosome Location
Chr.10: 123136051-123137731 on Build GRCh38
Transcripts
NM_001105574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6566364 c.609G>A p.A203A Chr.10: 123137266-123137266 on Build GRCh38
682933 c.574G>A p.A192T Chr.10: 123137231-123137231 on Build GRCh38
6837806 c.605C>T p.A202V Chr.10: 123137262-123137262 on Build GRCh38
6898769 c.699C>T p.V233V Chr.10: 123137356-123137356 on Build GRCh38
5549148 c.672G>A p.A224A Chr.10: 123137329-123137329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922245756 Chr.10: 123137192-123137192 on Build GRCh38
rs748087707 Chr.10: 123137204-123137204 on Build GRCh38
rs776114764 Chr.10: 123137233-123137233 on Build GRCh38
rs770573334 Chr.10: 123137230-123137230 on Build GRCh38
rs766586446 Chr.10: 123137334-123137334 on Build GRCh38
rs780213242 Chr.10: 123137257-123137257 on Build GRCh38
rs750592257 Chr.10: 123137254-123137254 on Build GRCh38
rs372769291 Chr.10: 123137361-123137361 on Build GRCh38
rs767824635 Chr.10: 123137253-123137253 on Build GRCh38
rs762291045 Chr.10: 123137309-123137309 on Build GRCh38
rs957233829 Chr.10: 123137281-123137281 on Build GRCh38
rs935097380 Chr.10: 123137196-123137196 on Build GRCh38
rs767761887 Chr.10: 123137314-123137314 on Build GRCh38
rs1026187707 Chr.10: 123137326-123137326 on Build GRCh38
rs757141423 Chr.10: 123137347-123137347 on Build GRCh38
rs1035706460 Chr.10: 123137266-123137266 on Build GRCh38
rs773080829 Chr.10: 123137220-123137220 on Build GRCh38
rs777814621 Chr.10: 123137338-123137338 on Build GRCh38
rs376174025 Chr.10: 123137276-123137276 on Build GRCh38
rs776042038 Chr.10: 123137242-123137242 on Build GRCh38
rs909225515 Chr.10: 123137336-123137338 on Build GRCh38
rs755890737 Chr.10: 123137261-123137261 on Build GRCh38
rs1052582464 Chr.10: 123137197-123137197 on Build GRCh38
rs750727543 Chr.10: 123137327-123137327 on Build GRCh38
rs769300113 Chr.10: 123137291-123137291 on Build GRCh38
rs376762346 Chr.10: 123137302-123137302 on Build GRCh38
rs755073796 Chr.10: 123137337-123137337 on Build GRCh38
rs760920188 Chr.10: 123137333-123137333 on Build GRCh38
rs971662635 Chr.10: 123137298-123137298 on Build GRCh38
rs45492702 Chr.10: 123137301-123137301 on Build GRCh38
rs1020115800 Chr.10: 123137295-123137295 on Build GRCh38
rs780954487 Chr.10: 123137282-123137282 on Build GRCh38
rs769112328 Chr.10: 123137306-123137306 on Build GRCh38
rs76168984 Chr.10: 123137362-123137362 on Build GRCh38
rs764458020 Chr.10: 123137247-123137247 on Build GRCh38
rs764711185 Chr.10: 123137239-123137239 on Build GRCh38
rs372099696 Chr.10: 123137383-123137383 on Build GRCh38
rs771994241 Chr.10: 123137217-123137217 on Build GRCh38
rs746859866 Chr.10: 123137274-123137274 on Build GRCh38
rs952817338 Chr.10: 123137350-123137350 on Build GRCh38
rs751437973 Chr.10: 123137346-123137346 on Build GRCh38
rs758357440 Chr.10: 123137259-123137259 on Build GRCh38
rs756236030 Chr.10: 123137186-123137186 on Build GRCh38
rs187759881 Chr.10: 123137226-123137226 on Build GRCh38
rs373545765 Chr.10: 123137236-123137236 on Build GRCh38
rs779572270 Chr.10: 123137386-123137386 on Build GRCh38
rs763385456 Chr.10: 123137245-123137245 on Build GRCh38
rs529176331 Chr.10: 123137255-123137255 on Build GRCh38
rs748107953 Chr.10: 123137258-123137258 on Build GRCh38
rs779044690 Chr.10: 123137195-123137195 on Build GRCh38
rs377194232 Chr.10: 123137368-123137368 on Build GRCh38
rs757453579 Chr.10: 123137251-123137251 on Build GRCh38
rs774693341 Chr.10: 123137307-123137307 on Build GRCh38
rs751903736 Chr.10: 123137250-123137250 on Build GRCh38

View Full Product Details