Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 91261129-91261372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HFM1
Entrez Gene ID
164045
Gene Name
HFM1, ATP dependent DNA helicase homolog
Gene Aliases
MER3, POF9, SEC63D1, Si-11, Si-11-6, helicase
UniGene
Hs.454818
Gene Chromosome Location
Chr.1: 91260766-91408008 on Build GRCh38
Transcripts
NM_001017975.4 XM_011540849.1 XM_011540850.2 XM_011540851.1 XM_011540852.2 XM_011540855.1 XM_011540857.1 XM_011540859.2 XM_017000490.1 XM_017000491.1 XM_017000492.1 XM_017000493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3689883 c.4295A>G p.D1432G Chr.1: 91261303-91261303 on Build GRCh38
5735527 c.4253A>T p.D1418V Chr.1: 91261345-91261345 on Build GRCh38
5677032 c.4290A>G p.I1430M Chr.1: 91261308-91261308 on Build GRCh38
3361049 c.4275G>A p.K1425K Chr.1: 91261323-91261323 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747292830 Chr.1: 91261265-91261265 on Build GRCh38
rs759873559 Chr.1: 91261280-91261283 on Build GRCh38
rs371512737 Chr.1: 91261315-91261315 on Build GRCh38
rs995291787 Chr.1: 91261247-91261247 on Build GRCh38
rs772238850 Chr.1: 91261259-91261259 on Build GRCh38
rs900925297 Chr.1: 91261269-91261269 on Build GRCh38
rs376608073 Chr.1: 91261317-91261317 on Build GRCh38
rs866734042 Chr.1: 91261208-91261208 on Build GRCh38
rs372715588 Chr.1: 91261180-91261180 on Build GRCh38
rs933823135 Chr.1: 91261191-91261191 on Build GRCh38
rs369770429 Chr.1: 91261344-91261344 on Build GRCh38
rs1050975979 Chr.1: 91261192-91261192 on Build GRCh38
rs774472209 Chr.1: 91261200-91261200 on Build GRCh38
rs1039839980 Chr.1: 91261245-91261245 on Build GRCh38
rs762588618 Chr.1: 91261324-91261328 on Build GRCh38
rs771417978 Chr.1: 91261272-91261272 on Build GRCh38
rs79868049 Chr.1: 91261194-91261194 on Build GRCh38
rs183715671 Chr.1: 91261193-91261193 on Build GRCh38
rs778122725 Chr.1: 91261262-91261262 on Build GRCh38
rs759959365 Chr.1: 91261300-91261300 on Build GRCh38
rs748140149 Chr.1: 91261244-91261244 on Build GRCh38
rs763642049 Chr.1: 91261338-91261338 on Build GRCh38
rs762338817 Chr.1: 91261322-91261322 on Build GRCh38
rs189998243 Chr.1: 91261298-91261298 on Build GRCh38
rs537146026 Chr.1: 91261293-91261293 on Build GRCh38
rs921062854 Chr.1: 91261178-91261178 on Build GRCh38
rs899203929 Chr.1: 91261206-91261206 on Build GRCh38

View Full Product Details