Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24781510-24781737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMNN
Entrez Gene ID
51053
Gene Name
geminin, DNA replication inhibitor
Gene Aliases
Gem, MGORS6
UniGene
Hs.234896
Gene Chromosome Location
Chr.6: 24774931-24786099 on Build GRCh38
Transcripts
NM_001251989.1 NM_001251990.1 NM_001251991.1 NM_015895.4 XM_005249159.1 XM_011514651.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4751372 c.178T>C p.S60P Chr.6: 24781525-24781525 on Build GRCh38
6346919 c.211G>T p.E71* Chr.6: 24781558-24781558 on Build GRCh38
740983 c.230A>G p.N77S Chr.6: 24781577-24781577 on Build GRCh38
5950124 c.256T>C p.F86L Chr.6: 24781603-24781603 on Build GRCh38
5184091 c.274+2T>C p.? Chr.6: 24781623-24781623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954425522 Chr.6: 24781686-24781686 on Build GRCh38
rs113023646 Chr.6: 24781600-24781600 on Build GRCh38
rs754589141 Chr.6: 24781583-24781583 on Build GRCh38
rs577488599 Chr.6: 24781613-24781613 on Build GRCh38
rs1035809642 Chr.6: 24781549-24781549 on Build GRCh38
rs776527320 Chr.6: 24781658-24781658 on Build GRCh38
rs1004696853 Chr.6: 24781592-24781592 on Build GRCh38
rs778546231 Chr.6: 24781593-24781593 on Build GRCh38
rs763108405 Chr.6: 24781540-24781540 on Build GRCh38
rs757821319 Chr.6: 24781579-24781579 on Build GRCh38
rs771605296 Chr.6: 24781625-24781625 on Build GRCh38
rs766774444 Chr.6: 24781703-24781703 on Build GRCh38
rs138954355 Chr.6: 24781581-24781581 on Build GRCh38
rs201417437 Chr.6: 24781633-24781633 on Build GRCh38
rs1029071851 Chr.6: 24781664-24781664 on Build GRCh38
rs369471914 Chr.6: 24781649-24781649 on Build GRCh38
rs370562633 Chr.6: 24781573-24781573 on Build GRCh38
rs373566295 Chr.6: 24781705-24781705 on Build GRCh38
rs989188728 Chr.6: 24781676-24781676 on Build GRCh38
rs764443451 Chr.6: 24781546-24781546 on Build GRCh38
rs777205317 Chr.6: 24781632-24781632 on Build GRCh38
rs575119675 Chr.6: 24781662-24781662 on Build GRCh38
rs543948104 Chr.6: 24781667-24781667 on Build GRCh38
rs781516962 Chr.6: 24781580-24781580 on Build GRCh38

View Full Product Details