Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94458269-94458542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD14B
Entrez Gene ID
84641
Gene Name
major facilitator superfamily domain containing 14B
Gene Aliases
HIATL1
UniGene
Hs.555996
Gene Chromosome Location
Chr.9: 94374327-94460920 on Build GRCh38
Transcripts
NM_032558.2 XM_006717305.3 XM_011519114.2 XM_017015217.1 XM_017015218.1 XM_017015219.1 XM_017015220.1 XM_017015221.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416148 c.1150G>T p.G384* Chr.9: 94458345-94458345 on Build GRCh38
2732873 c.1209G>A p.M403I Chr.9: 94458404-94458404 on Build GRCh38
6013537 c.1174C>T p.L392L Chr.9: 94458369-94458369 on Build GRCh38
1674634 c.1160G>A p.G387E Chr.9: 94458355-94458355 on Build GRCh38
6679802 c.1134C>A p.A378A Chr.9: 94458329-94458329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116615222 Chr.9: 94458364-94458364 on Build GRCh38
rs770838836 Chr.9: 94458473-94458473 on Build GRCh38
rs17851857 Chr.9: 94458411-94458411 on Build GRCh38
rs183889481 Chr.9: 94458359-94458359 on Build GRCh38
rs758547532 Chr.9: 94458346-94458346 on Build GRCh38
rs772989126 Chr.9: 94458383-94458383 on Build GRCh38
rs143110167 Chr.9: 94458339-94458339 on Build GRCh38
rs148361138 Chr.9: 94458311-94458311 on Build GRCh38
rs371824265 Chr.9: 94458503-94458503 on Build GRCh38
rs775253877 Chr.9: 94458488-94458488 on Build GRCh38
rs35026527 Chr.9: 94458321-94458321 on Build GRCh38
rs762684948 Chr.9: 94458501-94458504 on Build GRCh38
rs549758014 Chr.9: 94458306-94458306 on Build GRCh38
rs771918553 Chr.9: 94458485-94458485 on Build GRCh38
rs1027302940 Chr.9: 94458335-94458335 on Build GRCh38
rs779286212 Chr.9: 94458332-94458332 on Build GRCh38
rs754298657 Chr.9: 94458317-94458317 on Build GRCh38
rs752641750 Chr.9: 94458322-94458323 on Build GRCh38
rs755025692 Chr.9: 94458456-94458456 on Build GRCh38
rs996178522 Chr.9: 94458318-94458318 on Build GRCh38
rs774160325 Chr.9: 94458412-94458412 on Build GRCh38
rs778589915 Chr.9: 94458476-94458476 on Build GRCh38
rs984747622 Chr.9: 94458294-94458294 on Build GRCh38
rs374320089 Chr.9: 94458499-94458499 on Build GRCh38
rs764703511 Chr.9: 94458434-94458434 on Build GRCh38
rs1019767960 Chr.9: 94458502-94458502 on Build GRCh38
rs143357486 Chr.9: 94458458-94458458 on Build GRCh38
rs750741767 Chr.9: 94458450-94458450 on Build GRCh38
rs967975363 Chr.9: 94458465-94458465 on Build GRCh38
rs1018667032 Chr.9: 94458388-94458388 on Build GRCh38
rs757538168 Chr.9: 94458329-94458329 on Build GRCh38
rs62579752 Chr.9: 94458338-94458338 on Build GRCh38
rs776140320 Chr.9: 94458507-94458507 on Build GRCh38
rs775198616 Chr.9: 94458429-94458429 on Build GRCh38
rs763767917 Chr.9: 94458509-94458516 on Build GRCh38
rs764685460 Chr.9: 94458314-94458314 on Build GRCh38
rs754321909 Chr.9: 94458446-94458446 on Build GRCh38
rs767221285 Chr.9: 94458419-94458419 on Build GRCh38
rs199527138 Chr.9: 94458452-94458452 on Build GRCh38
rs201288774 Chr.9: 94458500-94458500 on Build GRCh38
rs769787347 Chr.9: 94458379-94458379 on Build GRCh38
rs747129360 Chr.9: 94458453-94458453 on Build GRCh38
rs760313152 Chr.9: 94458493-94458493 on Build GRCh38
rs757741069 Chr.9: 94458447-94458447 on Build GRCh38
rs758673136 Chr.9: 94458451-94458451 on Build GRCh38
rs11545563 Chr.9: 94458433-94458433 on Build GRCh38
rs745645917 Chr.9: 94458478-94458478 on Build GRCh38
rs767171892 Chr.9: 94458295-94458295 on Build GRCh38
rs777508991 Chr.9: 94458468-94458468 on Build GRCh38
rs979788664 Chr.9: 94458490-94458490 on Build GRCh38
rs770656557 Chr.9: 94458407-94458407 on Build GRCh38
rs139827537 Chr.9: 94458392-94458392 on Build GRCh38

View Full Product Details