Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173158645-173158910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP1
Entrez Gene ID
662
Gene Name
BCL2 interacting protein 1
Gene Aliases
NIP1, SEC20, TRG-8
UniGene
Hs.145726
Gene Chromosome Location
Chr.5: 173144442-173164387 on Build GRCh38
Transcripts
NM_001205.2 NM_013978.2 NM_013979.2 NM_013980.2 XM_011534638.1 XM_011534639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
737689 c.400A>C p.N134H Chr.5: 173158745-173158745 on Build GRCh38
3853758 c.481G>A p.G161R Chr.5: 173158826-173158826 on Build GRCh38
7086252 c.429C>G p.L143L Chr.5: 173158774-173158774 on Build GRCh38
1634156 c.410C>T p.S137L Chr.5: 173158755-173158755 on Build GRCh38
7185649 c.440T>C p.I147T Chr.5: 173158785-173158785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775280476 Chr.5: 173158735-173158735 on Build GRCh38
rs552081745 Chr.5: 173158684-173158684 on Build GRCh38
rs773628237 Chr.5: 173158710-173158710 on Build GRCh38
rs781529370 Chr.5: 173158707-173158707 on Build GRCh38
rs770588782 Chr.5: 173158714-173158714 on Build GRCh38
rs767108950 Chr.5: 173158725-173158725 on Build GRCh38
rs1025212234 Chr.5: 173158731-173158731 on Build GRCh38
rs370583969 Chr.5: 173158793-173158793 on Build GRCh38
rs890528070 Chr.5: 173158664-173158664 on Build GRCh38
rs781399578 Chr.5: 173158822-173158822 on Build GRCh38
rs970416574 Chr.5: 173158790-173158790 on Build GRCh38
rs1008961958 Chr.5: 173158671-173158671 on Build GRCh38
rs758700245 Chr.5: 173158696-173158696 on Build GRCh38
rs201405079 Chr.5: 173158724-173158724 on Build GRCh38
rs760735170 Chr.5: 173158748-173158748 on Build GRCh38
rs754999411 Chr.5: 173158814-173158814 on Build GRCh38
rs142359158 Chr.5: 173158792-173158792 on Build GRCh38
rs748746515 Chr.5: 173158708-173158708 on Build GRCh38
rs200835671 Chr.5: 173158874-173158874 on Build GRCh38
rs753077990 Chr.5: 173158824-173158824 on Build GRCh38
rs983482892 Chr.5: 173158791-173158791 on Build GRCh38
rs751871522 Chr.5: 173158802-173158802 on Build GRCh38
rs756656154 Chr.5: 173158826-173158826 on Build GRCh38
rs748879608 Chr.5: 173158712-173158712 on Build GRCh38
rs770048120 Chr.5: 173158709-173158709 on Build GRCh38
rs5745159 Chr.5: 173158704-173158704 on Build GRCh38
rs1032447286 Chr.5: 173158670-173158670 on Build GRCh38
rs536614812 Chr.5: 173158875-173158875 on Build GRCh38
rs987809153 Chr.5: 173158846-173158846 on Build GRCh38
rs773808162 Chr.5: 173158717-173158717 on Build GRCh38
rs766739783 Chr.5: 173158801-173158801 on Build GRCh38
rs780225753 Chr.5: 173158703-173158703 on Build GRCh38
rs147666879 Chr.5: 173158753-173158753 on Build GRCh38
rs764376980 Chr.5: 173158807-173158809 on Build GRCh38

View Full Product Details