Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43013731-43013996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI35
Entrez Gene ID
3430
Gene Name
interferon induced protein 35
Gene Aliases
IFP35
UniGene
Hs.632258
Gene Chromosome Location
Chr.17: 43006725-43014459 on Build GRCh38
Transcripts
NM_005533.4 XM_005257302.4 XM_017024584.1 XM_017024585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
215690 c.578G>A p.R193H Chr.17: 43013785-43013785 on Build GRCh38
7085243 c.600C>A p.F200L Chr.17: 43013807-43013807 on Build GRCh38
6418168 c.640T>C p.S214P Chr.17: 43013847-43013847 on Build GRCh38
5981228 c.635G>C p.R212T Chr.17: 43013842-43013842 on Build GRCh38
6683779 c.644C>T p.P215L Chr.17: 43013851-43013851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998516333 Chr.17: 43013875-43013875 on Build GRCh38
rs984191588 Chr.17: 43013922-43013922 on Build GRCh38
rs772115934 Chr.17: 43013838-43013838 on Build GRCh38
rs927995452 Chr.17: 43013937-43013937 on Build GRCh38
rs377160882 Chr.17: 43013909-43013909 on Build GRCh38
rs998449746 Chr.17: 43013891-43013891 on Build GRCh38
rs139500073 Chr.17: 43013947-43013947 on Build GRCh38
rs182267103 Chr.17: 43013927-43013927 on Build GRCh38
rs553080457 Chr.17: 43013923-43013923 on Build GRCh38
rs938559872 Chr.17: 43013942-43013942 on Build GRCh38
rs946662016 Chr.17: 43013840-43013840 on Build GRCh38
rs754226732 Chr.17: 43013885-43013885 on Build GRCh38
rs200811245 Chr.17: 43013851-43013851 on Build GRCh38
rs1013052001 Chr.17: 43013856-43013856 on Build GRCh38
rs752302821 Chr.17: 43013781-43013781 on Build GRCh38
rs766711749 Chr.17: 43013884-43013884 on Build GRCh38
rs780464597 Chr.17: 43013794-43013794 on Build GRCh38
rs972766292 Chr.17: 43013935-43013935 on Build GRCh38
rs370897840 Chr.17: 43013950-43013950 on Build GRCh38
rs897394863 Chr.17: 43013933-43013933 on Build GRCh38
rs777779265 Chr.17: 43013910-43013910 on Build GRCh38
rs761497733 Chr.17: 43013877-43013877 on Build GRCh38
rs745687267 Chr.17: 43013924-43013924 on Build GRCh38
rs934065641 Chr.17: 43013882-43013882 on Build GRCh38
rs763570551 Chr.17: 43013865-43013865 on Build GRCh38
rs199741958 Chr.17: 43013919-43013919 on Build GRCh38
rs13229 Chr.17: 43013861-43013861 on Build GRCh38
rs769873611 Chr.17: 43013853-43013853 on Build GRCh38
rs1017054603 Chr.17: 43013925-43013925 on Build GRCh38
rs756488054 Chr.17: 43013792-43013792 on Build GRCh38
rs370863270 Chr.17: 43013813-43013813 on Build GRCh38
rs550567872 Chr.17: 43013842-43013842 on Build GRCh38
rs755531972 Chr.17: 43013799-43013799 on Build GRCh38
rs761822573 Chr.17: 43013761-43013761 on Build GRCh38
rs754314991 Chr.17: 43013798-43013798 on Build GRCh38
rs748294501 Chr.17: 43013829-43013829 on Build GRCh38
rs569143868 Chr.17: 43013852-43013852 on Build GRCh38
rs779394952 Chr.17: 43013896-43013896 on Build GRCh38
rs146311145 Chr.17: 43013785-43013785 on Build GRCh38
rs992770210 Chr.17: 43013943-43013943 on Build GRCh38
rs528191120 Chr.17: 43013767-43013767 on Build GRCh38
rs760521218 Chr.17: 43013758-43013758 on Build GRCh38
rs756572425 Chr.17: 43013954-43013954 on Build GRCh38
rs747222139 Chr.17: 43013914-43013914 on Build GRCh38
rs372773230 Chr.17: 43013907-43013907 on Build GRCh38
rs751220759 Chr.17: 43013873-43013873 on Build GRCh38
rs201614442 Chr.17: 43013784-43013784 on Build GRCh38
rs200716829 Chr.17: 43013893-43013893 on Build GRCh38
rs373953401 Chr.17: 43013759-43013759 on Build GRCh38
rs765111811 Chr.17: 43013905-43013906 on Build GRCh38
rs764808276 Chr.17: 43013775-43013775 on Build GRCh38
rs537239237 Chr.17: 43013756-43013756 on Build GRCh38
rs372379979 Chr.17: 43013791-43013791 on Build GRCh38
rs776692577 Chr.17: 43013770-43013770 on Build GRCh38

View Full Product Details