Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114617105-114617378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
UniGene
Hs.522357
Gene Chromosome Location
Chr.9: 114603897-114646423 on Build GRCh38
Transcripts
NM_153045.3 XM_006716997.1 XM_011518350.1 XM_011518351.2 XM_011518352.1 XM_011518353.1 XM_011518354.1 XM_011518355.1 XM_017014429.1 XM_017014430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279499 c.106+5G>T p.? Chr.9: 114617306-114617306 on Build GRCh38
5711962 c.10G>A p.E4K Chr.9: 114617205-114617205 on Build GRCh38
4887090 c.106G>T p.E36* Chr.9: 114617301-114617301 on Build GRCh38
5835598 c.45_46insC p.S18fs*32 Chr.9: 114617240-114617241 on Build GRCh38
6048382 c.46delC p.S18fs*74 Chr.9: 114617241-114617241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748379717 Chr.9: 114617300-114617300 on Build GRCh38
rs757518784 Chr.9: 114617188-114617188 on Build GRCh38
rs746425907 Chr.9: 114617191-114617191 on Build GRCh38
rs773928523 Chr.9: 114617241-114617241 on Build GRCh38
rs748372095 Chr.9: 114617221-114617221 on Build GRCh38
rs775050812 Chr.9: 114617352-114617352 on Build GRCh38
rs957576158 Chr.9: 114617144-114617144 on Build GRCh38
rs779386076 Chr.9: 114617190-114617190 on Build GRCh38
rs780659537 Chr.9: 114617281-114617281 on Build GRCh38
rs930532391 Chr.9: 114617275-114617275 on Build GRCh38
rs758977140 Chr.9: 114617267-114617267 on Build GRCh38
rs906578639 Chr.9: 114617358-114617358 on Build GRCh38
rs766318080 Chr.9: 114617293-114617293 on Build GRCh38
rs373164511 Chr.9: 114617268-114617268 on Build GRCh38
rs747146259 Chr.9: 114617282-114617282 on Build GRCh38
rs147073169 Chr.9: 114617205-114617205 on Build GRCh38
rs543831980 Chr.9: 114617210-114617210 on Build GRCh38
rs776589076 Chr.9: 114617298-114617298 on Build GRCh38
rs765754148 Chr.9: 114617258-114617258 on Build GRCh38
rs772735157 Chr.9: 114617243-114617250 on Build GRCh38
rs939782717 Chr.9: 114617230-114617230 on Build GRCh38
rs1045877244 Chr.9: 114617218-114617218 on Build GRCh38
rs749071960 Chr.9: 114617177-114617177 on Build GRCh38
rs890938719 Chr.9: 114617266-114617266 on Build GRCh38
rs1007988867 Chr.9: 114617293-114617293 on Build GRCh38
rs772998450 Chr.9: 114617306-114617306 on Build GRCh38
rs772982068 Chr.9: 114617228-114617228 on Build GRCh38
rs762858729 Chr.9: 114617150-114617150 on Build GRCh38
rs781310001 Chr.9: 114617297-114617297 on Build GRCh38
rs747776872 Chr.9: 114617222-114617222 on Build GRCh38
rs760124032 Chr.9: 114617250-114617251 on Build GRCh38
rs767017553 Chr.9: 114617176-114617176 on Build GRCh38
rs755767755 Chr.9: 114617251-114617251 on Build GRCh38
rs762949101 Chr.9: 114617229-114617229 on Build GRCh38
rs751066996 Chr.9: 114617159-114617159 on Build GRCh38
rs754666454 Chr.9: 114617163-114617163 on Build GRCh38
rs750810487 Chr.9: 114617265-114617265 on Build GRCh38
rs145503787 Chr.9: 114617332-114617332 on Build GRCh38
rs925950462 Chr.9: 114617142-114617142 on Build GRCh38
rs376587972 Chr.9: 114617234-114617234 on Build GRCh38
rs200569038 Chr.9: 114617154-114617154 on Build GRCh38
rs769626474 Chr.9: 114617222-114617222 on Build GRCh38
rs759481659 Chr.9: 114617350-114617350 on Build GRCh38
rs771623478 Chr.9: 114617227-114617227 on Build GRCh38
rs369082056 Chr.9: 114617213-114617213 on Build GRCh38
rs758960646 Chr.9: 114617201-114617201 on Build GRCh38
rs1021227995 Chr.9: 114617299-114617299 on Build GRCh38
rs369771328 Chr.9: 114617249-114617249 on Build GRCh38
rs774131252 Chr.9: 114617218-114617219 on Build GRCh38
rs754249719 Chr.9: 114617187-114617187 on Build GRCh38
rs41278667 Chr.9: 114617309-114617309 on Build GRCh38
rs770874600 Chr.9: 114617326-114617326 on Build GRCh38
rs758918777 Chr.9: 114617246-114617246 on Build GRCh38
rs770003468 Chr.9: 114617302-114617302 on Build GRCh38
rs776679643 Chr.9: 114617214-114617214 on Build GRCh38
rs755052752 Chr.9: 114617295-114617295 on Build GRCh38

View Full Product Details