Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 50283242-50283469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXXC1
Entrez Gene ID
30827
Gene Name
CXXC finger protein 1
Gene Aliases
2410002I16Rik, 5830420C16Rik, CFP1, CGBP, HsT2645, PCCX1, PHF18, SPP1, ZCGPC1, hCGBP
UniGene
Hs.180933
Gene Chromosome Location
Chr.18: 50282343-50288322 on Build GRCh38
Transcripts
NM_001101654.1 NM_014593.3 XM_011525940.2 XM_011525941.2 XM_017025718.1 XM_017025719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5652732 c.1576G>T p.A526S Chr.18: 50283360-50283360 on Build GRCh38
2879312 c.1651G>A p.E551K Chr.18: 50283285-50283285 on Build GRCh38
4675724 c.1651G>T p.E551* Chr.18: 50283285-50283285 on Build GRCh38
4675725 c.1595A>G p.D532G Chr.18: 50283341-50283341 on Build GRCh38
1611273 c.1671+10delT p.? Chr.18: 50283255-50283255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761055392 Chr.18: 50283401-50283401 on Build GRCh38
rs756705322 Chr.18: 50283286-50283286 on Build GRCh38
rs754605356 Chr.18: 50283319-50283319 on Build GRCh38
rs777708237 Chr.18: 50283349-50283349 on Build GRCh38
rs760867157 Chr.18: 50283430-50283430 on Build GRCh38
rs183088108 Chr.18: 50283441-50283441 on Build GRCh38
rs754241327 Chr.18: 50283306-50283306 on Build GRCh38
rs34088160 Chr.18: 50283325-50283325 on Build GRCh38
rs767485856 Chr.18: 50283397-50283397 on Build GRCh38
rs142342206 Chr.18: 50283393-50283393 on Build GRCh38
rs909806286 Chr.18: 50283374-50283374 on Build GRCh38
rs970920151 Chr.18: 50283415-50283415 on Build GRCh38
rs113646896 Chr.18: 50283312-50283312 on Build GRCh38
rs770302767 Chr.18: 50283316-50283316 on Build GRCh38
rs202100050 Chr.18: 50283294-50283294 on Build GRCh38
rs780279436 Chr.18: 50283298-50283298 on Build GRCh38
rs768948215 Chr.18: 50283384-50283384 on Build GRCh38
rs747757661 Chr.18: 50283346-50283346 on Build GRCh38
rs377127282 Chr.18: 50283375-50283375 on Build GRCh38
rs565485071 Chr.18: 50283392-50283392 on Build GRCh38
rs768003078 Chr.18: 50283274-50283274 on Build GRCh38
rs369190736 Chr.18: 50283382-50283382 on Build GRCh38
rs372491324 Chr.18: 50283373-50283373 on Build GRCh38
rs980916990 Chr.18: 50283424-50283424 on Build GRCh38
rs200009297 Chr.18: 50283282-50283282 on Build GRCh38
rs190506121 Chr.18: 50283431-50283431 on Build GRCh38
rs368281485 Chr.18: 50283329-50283329 on Build GRCh38
rs918171801 Chr.18: 50283414-50283414 on Build GRCh38
rs150530383 Chr.18: 50283340-50283340 on Build GRCh38
rs766960100 Chr.18: 50283412-50283412 on Build GRCh38
rs992899998 Chr.18: 50283405-50283405 on Build GRCh38
rs747616387 Chr.18: 50283336-50283336 on Build GRCh38
rs776381264 Chr.18: 50283365-50283365 on Build GRCh38
rs916932625 Chr.18: 50283406-50283406 on Build GRCh38
rs901052062 Chr.18: 50283366-50283366 on Build GRCh38
rs908619789 Chr.18: 50283445-50283445 on Build GRCh38
rs968475164 Chr.18: 50283376-50283376 on Build GRCh38
rs949775531 Chr.18: 50283418-50283418 on Build GRCh38
rs750586999 Chr.18: 50283399-50283399 on Build GRCh38

View Full Product Details