Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 33597112-33597371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP26
Entrez Gene ID
78986
Gene Name
dual specificity phosphatase 26 (putative)
Gene Aliases
DSP-4, DUSP24, LDP-4, MKP-8, MKP8, NATA1, NEAP, SKRP3
UniGene
Hs.8719
Gene Chromosome Location
Chr.8: 33591330-33600106 on Build GRCh38
Transcripts
NM_001305115.1 NM_001305116.1 NM_024025.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3703397 c.221+1G>C p.? Chr.8: 33597294-33597294 on Build GRCh38
387886 c.180C>A p.N60K Chr.8: 33597336-33597336 on Build GRCh38
320036 c.164G>T p.G55V Chr.8: 33597352-33597352 on Build GRCh38
3996032 c.161C>A p.T54K Chr.8: 33597355-33597355 on Build GRCh38
2959242 c.192G>T p.E64D Chr.8: 33597324-33597324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117959564 Chr.8: 33597269-33597269 on Build GRCh38
rs768974445 Chr.8: 33597280-33597280 on Build GRCh38
rs763053747 Chr.8: 33597159-33597159 on Build GRCh38
rs755209158 Chr.8: 33597263-33597263 on Build GRCh38
rs200175837 Chr.8: 33597285-33597285 on Build GRCh38
rs774485490 Chr.8: 33597275-33597275 on Build GRCh38
rs747239034 Chr.8: 33597262-33597262 on Build GRCh38
rs73612670 Chr.8: 33597242-33597242 on Build GRCh38
rs757181813 Chr.8: 33597347-33597347 on Build GRCh38
rs921774621 Chr.8: 33597153-33597153 on Build GRCh38
rs758873163 Chr.8: 33597310-33597310 on Build GRCh38
rs754975205 Chr.8: 33597329-33597329 on Build GRCh38
rs758531240 Chr.8: 33597261-33597261 on Build GRCh38
rs551242446 Chr.8: 33597198-33597198 on Build GRCh38
rs369030554 Chr.8: 33597270-33597270 on Build GRCh38
rs749789957 Chr.8: 33597251-33597252 on Build GRCh38
rs776198250 Chr.8: 33597291-33597291 on Build GRCh38
rs112800969 Chr.8: 33597268-33597268 on Build GRCh38
rs974601117 Chr.8: 33597144-33597144 on Build GRCh38
rs559949748 Chr.8: 33597205-33597205 on Build GRCh38
rs745791396 Chr.8: 33597283-33597283 on Build GRCh38
rs1035090254 Chr.8: 33597150-33597150 on Build GRCh38
rs751685161 Chr.8: 33597327-33597327 on Build GRCh38
rs895305268 Chr.8: 33597299-33597299 on Build GRCh38
rs373673469 Chr.8: 33597226-33597226 on Build GRCh38
rs760054369 Chr.8: 33597286-33597286 on Build GRCh38
rs985107376 Chr.8: 33597313-33597313 on Build GRCh38
rs750874113 Chr.8: 33597260-33597260 on Build GRCh38
rs764345877 Chr.8: 33597264-33597264 on Build GRCh38
rs748365596 Chr.8: 33597345-33597345 on Build GRCh38
rs150109623 Chr.8: 33597284-33597284 on Build GRCh38
rs375557504 Chr.8: 33597307-33597307 on Build GRCh38
rs766421522 Chr.8: 33597326-33597326 on Build GRCh38
rs757576093 Chr.8: 33597245-33597245 on Build GRCh38
rs193214209 Chr.8: 33597330-33597330 on Build GRCh38
rs763836838 Chr.8: 33597288-33597288 on Build GRCh38
rs115545327 Chr.8: 33597303-33597303 on Build GRCh38
rs533125188 Chr.8: 33597163-33597163 on Build GRCh38
rs777993513 Chr.8: 33597168-33597168 on Build GRCh38
rs372186090 Chr.8: 33597302-33597302 on Build GRCh38
rs549089171 Chr.8: 33597256-33597256 on Build GRCh38
rs868348775 Chr.8: 33597164-33597164 on Build GRCh38

View Full Product Details