Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4747250-4747511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf71
Entrez Gene ID
146562
Gene Name
chromosome 16 open reading frame 71
Gene Aliases
-
UniGene
Hs.602738
Gene Chromosome Location
Chr.16: 4734288-4749396 on Build GRCh38
Transcripts
NM_139170.2 XM_005255144.3 XM_017022975.1 XM_017022976.1 XM_011522453.2

Gene Symbol
ZNF500
Entrez Gene ID
26048
Gene Name
zinc finger protein 500
Gene Aliases
ZKSCAN18, ZSCAN50
UniGene
Hs.513316
Gene Chromosome Location
Chr.16: 4745961-4767219 on Build GRCh38
Transcripts
NM_139170.2 XM_005255144.3 XM_017022975.1 XM_017022976.1 XM_011522453.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5527563 c.1334G>A p.G445E Chr.16: 4747396-4747396 on Build GRCh38
374204 c.1367G>A p.G456E Chr.16: 4747429-4747429 on Build GRCh38
1728536 c.1327C>T p.Q443* Chr.16: 4747389-4747389 on Build GRCh38
1563101 c.1312C>T p.Q438* Chr.16: 4747374-4747374 on Build GRCh38
1718201 c.1298G>A p.S433N Chr.16: 4747360-4747360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763254563 Chr.16: 4747341-4747341 on Build GRCh38
rs199642499 Chr.16: 4747405-4747405 on Build GRCh38
rs770178058 Chr.16: 4747331-4747331 on Build GRCh38
rs773664937 Chr.16: 4747340-4747340 on Build GRCh38
rs749742281 Chr.16: 4747414-4747414 on Build GRCh38
rs764746497 Chr.16: 4747433-4747433 on Build GRCh38
rs527654261 Chr.16: 4747387-4747387 on Build GRCh38
rs79295075 Chr.16: 4747406-4747406 on Build GRCh38
rs755290242 Chr.16: 4747441-4747441 on Build GRCh38
rs374196793 Chr.16: 4747476-4747476 on Build GRCh38
rs755235213 Chr.16: 4747404-4747404 on Build GRCh38
rs151153820 Chr.16: 4747436-4747436 on Build GRCh38
rs878906775 Chr.16: 4747413-4747413 on Build GRCh38
rs149468321 Chr.16: 4747440-4747440 on Build GRCh38
rs1014950377 Chr.16: 4747482-4747482 on Build GRCh38
rs758176380 Chr.16: 4747300-4747300 on Build GRCh38
rs780162180 Chr.16: 4747304-4747304 on Build GRCh38
rs377538294 Chr.16: 4747383-4747383 on Build GRCh38
rs35842369 Chr.16: 4747337-4747338 on Build GRCh38
rs370017246 Chr.16: 4747309-4747309 on Build GRCh38
rs373473822 Chr.16: 4747353-4747353 on Build GRCh38
rs200705043 Chr.16: 4747299-4747299 on Build GRCh38
rs148678041 Chr.16: 4747410-4747410 on Build GRCh38
rs750388784 Chr.16: 4747340-4747340 on Build GRCh38
rs971576231 Chr.16: 4747452-4747452 on Build GRCh38
rs756157506 Chr.16: 4747458-4747458 on Build GRCh38
rs771202054 Chr.16: 4747415-4747415 on Build GRCh38
rs753402412 Chr.16: 4747352-4747352 on Build GRCh38
rs547787494 Chr.16: 4747396-4747396 on Build GRCh38
rs118001246 Chr.16: 4747481-4747481 on Build GRCh38
rs200070828 Chr.16: 4747409-4747409 on Build GRCh38
rs201846778 Chr.16: 4747465-4747465 on Build GRCh38
rs370127789 Chr.16: 4747391-4747391 on Build GRCh38
rs765123928 Chr.16: 4747369-4747369 on Build GRCh38
rs376752805 Chr.16: 4747308-4747308 on Build GRCh38
rs758141364 Chr.16: 4747389-4747389 on Build GRCh38
rs142065357 Chr.16: 4747431-4747431 on Build GRCh38
rs375845445 Chr.16: 4747435-4747435 on Build GRCh38
rs750333138 Chr.16: 4747434-4747434 on Build GRCh38
rs758119561 Chr.16: 4747379-4747380 on Build GRCh38
rs952097479 Chr.16: 4747293-4747293 on Build GRCh38
rs776229179 Chr.16: 4747432-4747432 on Build GRCh38
rs763654265 Chr.16: 4747360-4747360 on Build GRCh38
rs554554378 Chr.16: 4747471-4747471 on Build GRCh38
rs772381788 Chr.16: 4747480-4747480 on Build GRCh38
rs748721284 Chr.16: 4747327-4747327 on Build GRCh38
rs765122035 Chr.16: 4747385-4747385 on Build GRCh38
rs118086912 Chr.16: 4747355-4747355 on Build GRCh38
rs771449884 Chr.16: 4747473-4747473 on Build GRCh38
rs774690583 Chr.16: 4747428-4747428 on Build GRCh38
rs901686269 Chr.16: 4747368-4747368 on Build GRCh38
rs17853375 Chr.16: 4747456-4747456 on Build GRCh38
rs746265671 Chr.16: 4747477-4747477 on Build GRCh38
rs749824660 Chr.16: 4747467-4747467 on Build GRCh38
rs962002216 Chr.16: 4747310-4747310 on Build GRCh38
rs1054886142 Chr.16: 4747328-4747328 on Build GRCh38
rs781595597 Chr.16: 4747455-4747455 on Build GRCh38
rs922514534 Chr.16: 4747349-4747349 on Build GRCh38

View Full Product Details