Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 79407059-79407306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GK2
Entrez Gene ID
2712
Gene Name
glycerol kinase 2
Gene Aliases
GKP2, GKTA
UniGene
Hs.98008
Gene Chromosome Location
Chr.4: 79406353-79408218 on Build GRCh38
Transcripts
NM_033214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369499 c.1118A>T p.E373V Chr.4: 79407083-79407083 on Build GRCh38
1058002 c.933G>T p.E311D Chr.4: 79407268-79407268 on Build GRCh38
1058000 c.1040G>T p.R347I Chr.4: 79407161-79407161 on Build GRCh38
2954982 c.958G>T p.G320C Chr.4: 79407243-79407243 on Build GRCh38
2954976 c.1091T>G p.F364C Chr.4: 79407110-79407110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866917714 Chr.4: 79407268-79407268 on Build GRCh38
rs764827524 Chr.4: 79407280-79407280 on Build GRCh38
rs773092299 Chr.4: 79407118-79407118 on Build GRCh38
rs7673905 Chr.4: 79407163-79407163 on Build GRCh38
rs1005241146 Chr.4: 79407084-79407084 on Build GRCh38
rs757517049 Chr.4: 79407160-79407160 on Build GRCh38
rs776671840 Chr.4: 79407254-79407254 on Build GRCh38
rs764765869 Chr.4: 79407144-79407144 on Build GRCh38
rs752444892 Chr.4: 79407145-79407145 on Build GRCh38
rs137991824 Chr.4: 79407109-79407109 on Build GRCh38
rs748431542 Chr.4: 79407215-79407215 on Build GRCh38
rs532948584 Chr.4: 79407124-79407124 on Build GRCh38
rs200729434 Chr.4: 79407229-79407229 on Build GRCh38
rs773002678 Chr.4: 79407251-79407251 on Build GRCh38
rs1015337805 Chr.4: 79407088-79407088 on Build GRCh38
rs753333072 Chr.4: 79407128-79407128 on Build GRCh38
rs749293008 Chr.4: 79407197-79407197 on Build GRCh38
rs374732833 Chr.4: 79407181-79407181 on Build GRCh38
rs772393135 Chr.4: 79407221-79407226 on Build GRCh38
rs765816887 Chr.4: 79407127-79407127 on Build GRCh38
rs774670514 Chr.4: 79407098-79407098 on Build GRCh38
rs768751739 Chr.4: 79407097-79407097 on Build GRCh38
rs988949687 Chr.4: 79407091-79407091 on Build GRCh38
rs759152060 Chr.4: 79407138-79407138 on Build GRCh38
rs371750502 Chr.4: 79407135-79407135 on Build GRCh38
rs549791960 Chr.4: 79407202-79407202 on Build GRCh38
rs747376869 Chr.4: 79407086-79407086 on Build GRCh38
rs749473478 Chr.4: 79407096-79407096 on Build GRCh38
rs149474069 Chr.4: 79407212-79407212 on Build GRCh38
rs781716007 Chr.4: 79407167-79407167 on Build GRCh38
rs947564396 Chr.4: 79407155-79407155 on Build GRCh38
rs937575078 Chr.4: 79407224-79407224 on Build GRCh38
rs746864179 Chr.4: 79407253-79407253 on Build GRCh38
rs758948027 Chr.4: 79407265-79407265 on Build GRCh38
rs761488569 Chr.4: 79407107-79407107 on Build GRCh38
rs199938688 Chr.4: 79407223-79407224 on Build GRCh38
rs755276822 Chr.4: 79407179-79407179 on Build GRCh38
rs780767905 Chr.4: 79407185-79407185 on Build GRCh38

View Full Product Details