Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 40933498-40933761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN1
Entrez Gene ID
1272
Gene Name
contactin 1
Gene Aliases
F3, GP135, MYPCN
UniGene
Hs.143434
Gene Chromosome Location
Chr.12: 40692439-41072412 on Build GRCh38
Transcripts
NM_001256063.1 NM_001256064.1 NM_001843.3 NM_175038.2 XM_005268651.2 XM_006719241.2 XM_011537926.2 XM_011537927.2 XM_017018821.1 XM_017018822.1 XM_017018823.1 XM_017018824.1 XM_017018825.1 XM_017018826.1 XM_017018827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4207049 c.793G>T p.A265S Chr.12: 40933550-40933550 on Build GRCh38
1628585 c.751G>T p.V251L Chr.12: 40933508-40933508 on Build GRCh38
6072348 c.755A>G p.Y252C Chr.12: 40933512-40933512 on Build GRCh38
6631702 c.752T>C p.V251A Chr.12: 40933509-40933509 on Build GRCh38
3792572 c.801A>T p.G267G Chr.12: 40933558-40933558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778411720 Chr.12: 40933706-40933706 on Build GRCh38
rs761240540 Chr.12: 40933598-40933598 on Build GRCh38
rs937462249 Chr.12: 40933698-40933698 on Build GRCh38
rs778449042 Chr.12: 40933581-40933581 on Build GRCh38
rs200759494 Chr.12: 40933675-40933675 on Build GRCh38
rs747859026 Chr.12: 40933708-40933708 on Build GRCh38
rs554314947 Chr.12: 40933612-40933612 on Build GRCh38
rs375708810 Chr.12: 40933676-40933676 on Build GRCh38
rs572715662 Chr.12: 40933616-40933616 on Build GRCh38
rs756134091 Chr.12: 40933680-40933680 on Build GRCh38
rs779778967 Chr.12: 40933543-40933543 on Build GRCh38
rs56323985 Chr.12: 40933625-40933625 on Build GRCh38
rs796927032 Chr.12: 40933624-40933625 on Build GRCh38
rs201607834 Chr.12: 40933720-40933720 on Build GRCh38
rs771622826 Chr.12: 40933593-40933593 on Build GRCh38
rs55976072 Chr.12: 40933624-40933624 on Build GRCh38
rs771256157 Chr.12: 40933603-40933603 on Build GRCh38
rs771359796 Chr.12: 40933721-40933721 on Build GRCh38
rs747626974 Chr.12: 40933589-40933589 on Build GRCh38
rs199622338 Chr.12: 40933701-40933701 on Build GRCh38
rs369343021 Chr.12: 40933660-40933660 on Build GRCh38
rs746184282 Chr.12: 40933730-40933730 on Build GRCh38
rs773699933 Chr.12: 40933595-40933595 on Build GRCh38
rs56172264 Chr.12: 40933689-40933689 on Build GRCh38
rs748913992 Chr.12: 40933574-40933574 on Build GRCh38
rs779849920 Chr.12: 40933685-40933685 on Build GRCh38
rs1042788910 Chr.12: 40933661-40933661 on Build GRCh38
rs144047921 Chr.12: 40933719-40933719 on Build GRCh38
rs957638697 Chr.12: 40933630-40933630 on Build GRCh38
rs374200408 Chr.12: 40933714-40933714 on Build GRCh38
rs200611219 Chr.12: 40933625-40933625 on Build GRCh38
rs199647238 Chr.12: 40933623-40933623 on Build GRCh38
rs967383840 Chr.12: 40933682-40933682 on Build GRCh38
rs139270901 Chr.12: 40933530-40933530 on Build GRCh38
rs764970741 Chr.12: 40933683-40933683 on Build GRCh38
rs1051869143 Chr.12: 40933623-40933623 on Build GRCh38
rs374647402 Chr.12: 40933662-40933662 on Build GRCh38
rs1036256810 Chr.12: 40933735-40933735 on Build GRCh38
rs928683884 Chr.12: 40933697-40933697 on Build GRCh38
rs375392779 Chr.12: 40933575-40933575 on Build GRCh38
rs200145508 Chr.12: 40933670-40933670 on Build GRCh38
rs776868385 Chr.12: 40933610-40933610 on Build GRCh38

View Full Product Details