Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101370775-101371030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAAT
Entrez Gene ID
570
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Gene Aliases
BACAT, BAT
UniGene
Hs.284712
Gene Chromosome Location
Chr.9: 101360417-101385005 on Build GRCh38
Transcripts
NM_001127610.1 NM_001701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652354 c.416G>A p.R139Q Chr.9: 101370989-101370989 on Build GRCh38
6013559 c.460C>T p.P154S Chr.9: 101370945-101370945 on Build GRCh38
3652353 c.434G>A p.G145D Chr.9: 101370971-101370971 on Build GRCh38
1283619 c.400G>T p.A134S Chr.9: 101371005-101371005 on Build GRCh38
1674676 c.427C>T p.R143* Chr.9: 101370978-101370978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041284283 Chr.9: 101370925-101370925 on Build GRCh38
rs1033982824 Chr.9: 101370857-101370857 on Build GRCh38
rs779384536 Chr.9: 101370948-101370948 on Build GRCh38
rs773128969 Chr.9: 101370989-101370989 on Build GRCh38
rs746186421 Chr.9: 101370952-101370952 on Build GRCh38
rs968594726 Chr.9: 101370871-101370871 on Build GRCh38
rs772820138 Chr.9: 101370949-101370950 on Build GRCh38
rs992742029 Chr.9: 101370872-101370872 on Build GRCh38
rs377581251 Chr.9: 101370902-101370902 on Build GRCh38
rs866176840 Chr.9: 101370945-101370945 on Build GRCh38
rs775846552 Chr.9: 101370959-101370959 on Build GRCh38
rs542081749 Chr.9: 101370819-101370819 on Build GRCh38
rs757422184 Chr.9: 101370943-101370943 on Build GRCh38
rs942509456 Chr.9: 101370914-101370914 on Build GRCh38
rs988080396 Chr.9: 101370829-101370829 on Build GRCh38
rs201427312 Chr.9: 101370944-101370944 on Build GRCh38
rs973237287 Chr.9: 101370961-101370961 on Build GRCh38
rs1014788984 Chr.9: 101370881-101370881 on Build GRCh38
rs758946279 Chr.9: 101370921-101370921 on Build GRCh38
rs758405554 Chr.9: 101370901-101370901 on Build GRCh38
rs756202882 Chr.9: 101370930-101370930 on Build GRCh38
rs144591246 Chr.9: 101370996-101370996 on Build GRCh38
rs772702641 Chr.9: 101370953-101370953 on Build GRCh38
rs986094615 Chr.9: 101370981-101370981 on Build GRCh38
rs752835678 Chr.9: 101370904-101370904 on Build GRCh38
rs915267575 Chr.9: 101370866-101370866 on Build GRCh38
rs751506096 Chr.9: 101370997-101370997 on Build GRCh38
rs865961812 Chr.9: 101370873-101370873 on Build GRCh38
rs140848255 Chr.9: 101370993-101370993 on Build GRCh38
rs762646933 Chr.9: 101370990-101370990 on Build GRCh38
rs749544775 Chr.9: 101370942-101370942 on Build GRCh38
rs764182372 Chr.9: 101371006-101371006 on Build GRCh38
rs776238355 Chr.9: 101370965-101370965 on Build GRCh38
rs772876970 Chr.9: 101370968-101370968 on Build GRCh38
rs978736641 Chr.9: 101370879-101370879 on Build GRCh38
rs138613717 Chr.9: 101371002-101371002 on Build GRCh38
rs904250423 Chr.9: 101370826-101370826 on Build GRCh38
rs747460084 Chr.9: 101370962-101370962 on Build GRCh38
rs777764682 Chr.9: 101370931-101370931 on Build GRCh38
rs764187428 Chr.9: 101370963-101370963 on Build GRCh38
rs764771610 Chr.9: 101370969-101370969 on Build GRCh38
rs953427538 Chr.9: 101370977-101370977 on Build GRCh38

View Full Product Details