Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112265627-112265900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1AR
Entrez Gene ID
55435
Gene Name
adaptor related protein complex 1 associated regulatory protein
Gene Aliases
2C18, C4orf16, GBAR, PRO0971, gamma-BAR
UniGene
Hs.435991
Gene Chromosome Location
Chr.4: 112231727-112273110 on Build GRCh38
Transcripts
NM_001128426.2 NM_018569.5 NR_136622.1 XM_017008378.1 XM_017008379.1 XM_017008380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
288317 c.463G>A p.E155K Chr.4: 112265756-112265756 on Build GRCh38
1426390 c.462C>T p.F154F Chr.4: 112265755-112265755 on Build GRCh38
1050091 c.506G>A p.R169Q Chr.4: 112265799-112265799 on Build GRCh38
4662198 c.441T>C p.S147S Chr.4: 112265734-112265734 on Build GRCh38
6365783 c.499G>A p.V167I Chr.4: 112265792-112265792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776007372 Chr.4: 112265720-112265720 on Build GRCh38
rs373307429 Chr.4: 112265761-112265761 on Build GRCh38
rs774420435 Chr.4: 112265705-112265705 on Build GRCh38
rs765769313 Chr.4: 112265844-112265844 on Build GRCh38
rs376213545 Chr.4: 112265775-112265775 on Build GRCh38
rs746050291 Chr.4: 112265799-112265799 on Build GRCh38
rs779198486 Chr.4: 112265798-112265798 on Build GRCh38
rs749440664 Chr.4: 112265784-112265784 on Build GRCh38
rs746242999 Chr.4: 112265711-112265711 on Build GRCh38
rs368520661 Chr.4: 112265848-112265848 on Build GRCh38
rs762359168 Chr.4: 112265741-112265741 on Build GRCh38
rs568973796 Chr.4: 112265818-112265818 on Build GRCh38
rs751862110 Chr.4: 112265829-112265829 on Build GRCh38
rs775874218 Chr.4: 112265822-112265822 on Build GRCh38
rs770019005 Chr.4: 112265684-112265684 on Build GRCh38
rs758206805 Chr.4: 112265801-112265803 on Build GRCh38
rs777954465 Chr.4: 112265689-112265689 on Build GRCh38
rs138136155 Chr.4: 112265693-112265693 on Build GRCh38
rs759315206 Chr.4: 112265859-112265859 on Build GRCh38
rs891350206 Chr.4: 112265672-112265672 on Build GRCh38
rs1052459694 Chr.4: 112265671-112265671 on Build GRCh38
rs767376223 Chr.4: 112265758-112265758 on Build GRCh38
rs772845682 Chr.4: 112265739-112265739 on Build GRCh38
rs747370622 Chr.4: 112265824-112265824 on Build GRCh38
rs562251972 Chr.4: 112265731-112265731 on Build GRCh38
rs769949052 Chr.4: 112265719-112265719 on Build GRCh38
rs774173993 Chr.4: 112265856-112265856 on Build GRCh38
rs776947727 Chr.4: 112265830-112265830 on Build GRCh38
rs756156454 Chr.4: 112265768-112265768 on Build GRCh38
rs61734509 Chr.4: 112265755-112265755 on Build GRCh38
rs749292948 Chr.4: 112265691-112265691 on Build GRCh38
rs1049148248 Chr.4: 112265656-112265656 on Build GRCh38
rs754855886 Chr.4: 112265756-112265756 on Build GRCh38
rs899997263 Chr.4: 112265742-112265742 on Build GRCh38
rs761121554 Chr.4: 112265725-112265725 on Build GRCh38
rs539529426 Chr.4: 112265840-112265840 on Build GRCh38
rs1031760438 Chr.4: 112265843-112265843 on Build GRCh38
rs550579652 Chr.4: 112265787-112265787 on Build GRCh38
rs989875378 Chr.4: 112265860-112265860 on Build GRCh38
rs1027801658 Chr.4: 112265785-112265785 on Build GRCh38

View Full Product Details