Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 37139289-37139536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf42
Entrez Gene ID
65250
Gene Name
chromosome 5 open reading frame 42
Gene Aliases
Hug, JBTS17, OFD6
UniGene
Hs.586199
Gene Chromosome Location
Chr.5: 37063928-37249428 on Build GRCh38
Transcripts
NM_023073.3 XM_005248345.3 XM_005248346.3 XM_005248347.3 XM_005248349.3 XM_005248350.3 XM_006714491.3 XM_011514085.2 XM_011514086.2 XM_011514087.2 XM_011514088.2 XM_011514089.2 XM_011514090.2 XM_011514092.2 XM_011514094.2 XM_017009760.1 XM_017009761.1 XM_017009762.1 XM_017009763.1 XM_017009764.1 XM_017009765.1 XM_017009766.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6490689 c.8471-3delT p.? Chr.5: 37139373-37139373 on Build GRCh38
6201097 c.8501+9C>T p.? Chr.5: 37139331-37139331 on Build GRCh38
6470320 c.5179G>A p.D1727N Chr.5: 37139356-37139356 on Build GRCh38
3686317 c.8471-4_8471-3delTT p.? Chr.5: 37139373-37139374 on Build GRCh38
6201098 c.5195+9C>T p.? Chr.5: 37139331-37139331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773760998 Chr.5: 37139409-37139409 on Build GRCh38
rs775583113 Chr.5: 37139464-37139464 on Build GRCh38
rs984335002 Chr.5: 37139422-37139422 on Build GRCh38
rs1033912985 Chr.5: 37139362-37139362 on Build GRCh38
rs971750121 Chr.5: 37139482-37139482 on Build GRCh38
rs529033527 Chr.5: 37139406-37139406 on Build GRCh38
rs1015709306 Chr.5: 37139430-37139430 on Build GRCh38
rs991518920 Chr.5: 37139368-37139368 on Build GRCh38
rs34646696 Chr.5: 37139373-37139373 on Build GRCh38
rs755250685 Chr.5: 37139406-37139406 on Build GRCh38
rs760294066 Chr.5: 37139327-37139328 on Build GRCh38
rs763532957 Chr.5: 37139372-37139373 on Build GRCh38
rs749464494 Chr.5: 37139399-37139399 on Build GRCh38
rs757083511 Chr.5: 37139344-37139344 on Build GRCh38
rs761088183 Chr.5: 37139411-37139411 on Build GRCh38
rs899527120 Chr.5: 37139489-37139489 on Build GRCh38
rs750386274 Chr.5: 37139380-37139380 on Build GRCh38
rs938657101 Chr.5: 37139334-37139334 on Build GRCh38
rs766732302 Chr.5: 37139373-37139374 on Build GRCh38
rs568268742 Chr.5: 37139393-37139393 on Build GRCh38
rs368765405 Chr.5: 37139385-37139385 on Build GRCh38
rs772618124 Chr.5: 37139408-37139408 on Build GRCh38
rs779383793 Chr.5: 37139402-37139402 on Build GRCh38
rs368241148 Chr.5: 37139503-37139503 on Build GRCh38
rs751987460 Chr.5: 37139405-37139406 on Build GRCh38
rs922949951 Chr.5: 37139495-37139495 on Build GRCh38
rs763664569 Chr.5: 37139328-37139332 on Build GRCh38
rs372742239 Chr.5: 37139325-37139325 on Build GRCh38
rs929491713 Chr.5: 37139400-37139400 on Build GRCh38
rs376655170 Chr.5: 37139335-37139335 on Build GRCh38
rs897952265 Chr.5: 37139395-37139395 on Build GRCh38
rs1017918079 Chr.5: 37139455-37139455 on Build GRCh38
rs547301109 Chr.5: 37139498-37139498 on Build GRCh38
rs139793397 Chr.5: 37139348-37139348 on Build GRCh38

View Full Product Details