Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155085146-155085413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA3
Entrez Gene ID
1944
Gene Name
ephrin A3
Gene Aliases
EFL2, EPLG3, Ehk1-L, LERK3
UniGene
Hs.516656
Gene Chromosome Location
Chr.1: 155078872-155087538 on Build GRCh38
Transcripts
NM_004952.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6507831 c.227G>A p.G76E Chr.1: 155085189-155085189 on Build GRCh38
6533902 c.400C>G p.L134V Chr.1: 155085362-155085362 on Build GRCh38
5643905 c.405C>T p.G135G Chr.1: 155085367-155085367 on Build GRCh38
302116 c.277C>T p.R93C Chr.1: 155085239-155085239 on Build GRCh38
6652065 c.325G>A p.E109K Chr.1: 155085287-155085287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986928720 Chr.1: 155085244-155085244 on Build GRCh38
rs774013507 Chr.1: 155085207-155085207 on Build GRCh38
rs936150386 Chr.1: 155085296-155085296 on Build GRCh38
rs748571354 Chr.1: 155085196-155085196 on Build GRCh38
rs778291971 Chr.1: 155085280-155085280 on Build GRCh38
rs755180780 Chr.1: 155085253-155085253 on Build GRCh38
rs781551302 Chr.1: 155085183-155085183 on Build GRCh38
rs896843413 Chr.1: 155085178-155085189 on Build GRCh38
rs900252843 Chr.1: 155085271-155085271 on Build GRCh38
rs978312203 Chr.1: 155085371-155085371 on Build GRCh38
rs1039160890 Chr.1: 155085238-155085238 on Build GRCh38
rs767358183 Chr.1: 155085215-155085215 on Build GRCh38
rs371183568 Chr.1: 155085302-155085302 on Build GRCh38
rs748483095 Chr.1: 155085274-155085274 on Build GRCh38
rs77832800 Chr.1: 155085361-155085361 on Build GRCh38
rs770370218 Chr.1: 155085205-155085205 on Build GRCh38
rs139404791 Chr.1: 155085173-155085173 on Build GRCh38
rs749790807 Chr.1: 155085298-155085298 on Build GRCh38
rs773029045 Chr.1: 155085178-155085181 on Build GRCh38
rs71576043 Chr.1: 155085174-155085175 on Build GRCh38
rs201029831 Chr.1: 155085201-155085201 on Build GRCh38
rs759262433 Chr.1: 155085213-155085213 on Build GRCh38
rs770331893 Chr.1: 155085198-155085198 on Build GRCh38
rs867240605 Chr.1: 155085192-155085192 on Build GRCh38
rs138967579 Chr.1: 155085367-155085367 on Build GRCh38
rs891994295 Chr.1: 155085346-155085346 on Build GRCh38
rs142225795 Chr.1: 155085230-155085230 on Build GRCh38
rs907188438 Chr.1: 155085379-155085379 on Build GRCh38
rs1054567261 Chr.1: 155085345-155085345 on Build GRCh38
rs766378281 Chr.1: 155085237-155085237 on Build GRCh38
rs747266587 Chr.1: 155085179-155085179 on Build GRCh38
rs751723541 Chr.1: 155085250-155085250 on Build GRCh38
rs750509227 Chr.1: 155085169-155085169 on Build GRCh38
rs140391461 Chr.1: 155085283-155085283 on Build GRCh38
rs781461826 Chr.1: 155085254-155085254 on Build GRCh38
rs758469116 Chr.1: 155085235-155085235 on Build GRCh38
rs1531478 Chr.1: 155085204-155085204 on Build GRCh38
rs954288179 Chr.1: 155085206-155085206 on Build GRCh38
rs140459162 Chr.1: 155085229-155085229 on Build GRCh38
rs145094848 Chr.1: 155085174-155085174 on Build GRCh38

View Full Product Details