Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 101875087-101875352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF14
Entrez Gene ID
2259
Gene Name
fibroblast growth factor 14
Gene Aliases
FGF-14, FHF-4, FHF4, SCA27
UniGene
Hs.508616
Gene Chromosome Location
Chr.13: 101720855-102402428 on Build GRCh38
Transcripts
NM_001321931.1 NM_001321932.1 NM_001321933.1 NM_001321934.1 NM_001321935.1 NM_001321936.1 NM_001321937.1 NM_001321938.1 NM_001321939.1 NM_001321940.1 NM_001321941.1 NM_001321942.1 NM_001321943.1 NM_001321944.1 NM_001321945.1 NM_001321946.1 NM_001321947.1 NM_001321948.1 NM_001321949.1 NM_004115.3 NM_175929.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793088 c.226T>C p.Y76H Chr.13: 101875264-101875264 on Build GRCh38
6663507 c.317C>A p.S106Y Chr.13: 101875188-101875188 on Build GRCh38
3793087 c.241T>C p.Y81H Chr.13: 101875264-101875264 on Build GRCh38
6251556 c.304+9C>A p.? Chr.13: 101875177-101875177 on Build GRCh38
3955365 c.261C>G p.P87P Chr.13: 101875229-101875229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374010824 Chr.13: 101875293-101875293 on Build GRCh38
rs1033392710 Chr.13: 101875271-101875271 on Build GRCh38
rs772935086 Chr.13: 101875228-101875228 on Build GRCh38
rs778738810 Chr.13: 101875156-101875156 on Build GRCh38
rs776246675 Chr.13: 101875233-101875233 on Build GRCh38
rs775032992 Chr.13: 101875257-101875257 on Build GRCh38
rs758950207 Chr.13: 101875234-101875234 on Build GRCh38
rs532999301 Chr.13: 101875311-101875311 on Build GRCh38
rs901698390 Chr.13: 101875153-101875153 on Build GRCh38
rs934726392 Chr.13: 101875314-101875314 on Build GRCh38
rs780245199 Chr.13: 101875143-101875143 on Build GRCh38
rs756389168 Chr.13: 101875140-101875140 on Build GRCh38
rs757210660 Chr.13: 101875229-101875229 on Build GRCh38
rs372625840 Chr.13: 101875147-101875147 on Build GRCh38
rs544949865 Chr.13: 101875256-101875256 on Build GRCh38
rs367892182 Chr.13: 101875316-101875316 on Build GRCh38
rs754041595 Chr.13: 101875318-101875318 on Build GRCh38
rs370000987 Chr.13: 101875214-101875214 on Build GRCh38
rs771857870 Chr.13: 101875216-101875216 on Build GRCh38
rs750810333 Chr.13: 101875310-101875310 on Build GRCh38
rs138051469 Chr.13: 101875230-101875230 on Build GRCh38
rs574124501 Chr.13: 101875217-101875217 on Build GRCh38
rs755044357 Chr.13: 101875321-101875321 on Build GRCh38
rs979512689 Chr.13: 101875224-101875224 on Build GRCh38
rs778895882 Chr.13: 101875325-101875325 on Build GRCh38
rs760399220 Chr.13: 101875146-101875149 on Build GRCh38
rs754992081 Chr.13: 101875154-101875154 on Build GRCh38
rs768049427 Chr.13: 101875305-101875305 on Build GRCh38
rs746620650 Chr.13: 101875173-101875173 on Build GRCh38

View Full Product Details