Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 145805626-145805887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO30
Entrez Gene ID
84085
Gene Name
F-box protein 30
Gene Aliases
Fbx30
UniGene
Hs.421095
Gene Chromosome Location
Chr.6: 145798135-145864409 on Build GRCh38
Transcripts
NM_032145.4 XM_005267159.3 XM_011536177.2 XM_017011353.1 NR_038246.1

Gene Symbol
LOC100507557
Entrez Gene ID
100507557
Gene Name
uncharacterized LOC100507557
Gene Aliases
-
UniGene
Hs.124537
Gene Chromosome Location
Chr.6: 145734869-145886585 on Build GRCh38
Transcripts
NM_032145.4 XM_005267159.3 XM_011536177.2 XM_017011353.1 NR_038246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924612 c.529G>C p.E177Q Chr.6: 145805877-145805877 on Build GRCh38
360249 c.765A>G p.T255T Chr.6: 145805641-145805641 on Build GRCh38
7077224 c.614G>C p.R205T Chr.6: 145805792-145805792 on Build GRCh38
4607933 c.764C>A p.T255K Chr.6: 145805642-145805642 on Build GRCh38
5869470 c.581C>T p.A194V Chr.6: 145805825-145805825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552414580 Chr.6: 145805743-145805743 on Build GRCh38
rs775047081 Chr.6: 145805859-145805859 on Build GRCh38
rs766539534 Chr.6: 145805669-145805669 on Build GRCh38
rs747579932 Chr.6: 145805741-145805741 on Build GRCh38
rs918174400 Chr.6: 145805736-145805736 on Build GRCh38
rs921758882 Chr.6: 145805834-145805834 on Build GRCh38
rs752543124 Chr.6: 145805701-145805701 on Build GRCh38
rs777504125 Chr.6: 145805716-145805716 on Build GRCh38
rs767488636 Chr.6: 145805686-145805686 on Build GRCh38
rs373306995 Chr.6: 145805799-145805799 on Build GRCh38
rs1039157313 Chr.6: 145805805-145805805 on Build GRCh38
rs138185905 Chr.6: 145805681-145805681 on Build GRCh38
rs143268192 Chr.6: 145805835-145805835 on Build GRCh38
rs555174655 Chr.6: 145805861-145805861 on Build GRCh38
rs774571445 Chr.6: 145805792-145805792 on Build GRCh38
rs781490990 Chr.6: 145805751-145805751 on Build GRCh38
rs534890951 Chr.6: 145805850-145805850 on Build GRCh38
rs752530228 Chr.6: 145805815-145805815 on Build GRCh38
rs763128781 Chr.6: 145805667-145805667 on Build GRCh38
rs200388937 Chr.6: 145805734-145805734 on Build GRCh38
rs945275222 Chr.6: 145805650-145805650 on Build GRCh38
rs760550203 Chr.6: 145805824-145805824 on Build GRCh38
rs767576595 Chr.6: 145805806-145805806 on Build GRCh38
rs748566272 Chr.6: 145805756-145805756 on Build GRCh38
rs532426956 Chr.6: 145805735-145805735 on Build GRCh38
rs201026665 Chr.6: 145805853-145805853 on Build GRCh38
rs771197123 Chr.6: 145805785-145805785 on Build GRCh38
rs187731447 Chr.6: 145805848-145805848 on Build GRCh38
rs149566037 Chr.6: 145805713-145805713 on Build GRCh38
rs755807101 Chr.6: 145805706-145805706 on Build GRCh38
rs773254998 Chr.6: 145805662-145805662 on Build GRCh38
rs753400919 Chr.6: 145805722-145805722 on Build GRCh38
rs770010359 Chr.6: 145805757-145805757 on Build GRCh38
rs773525965 Chr.6: 145805764-145805764 on Build GRCh38
rs558883058 Chr.6: 145805771-145805771 on Build GRCh38
rs770064255 Chr.6: 145805658-145805658 on Build GRCh38
rs925334307 Chr.6: 145805773-145805773 on Build GRCh38

View Full Product Details